Skip to Content
Merck

Skip To

39625

NTA - Atto 488

BioReagent, suitable for fluorescence, ≥80% (sum of isomers, HPCE)

Synonym(s):

Atto 488 - Nitrilotriacetic acid, complexed to Ni2+ - ion, Nα,Nα-bis(carboxymethyl)-L-lysine, Nickel(II) complex

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12171501
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥80% (sum of isomers, HPCE)

form

solid

fluorescence

λex 501 nm; λem 523 nm in PBS with 0.08% sodium azide

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

General description

find more information here

Application

for the fluorescence detection of HIS-tagged proteins

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
941590217541698
suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

fluorescence

λex 501 nm; λem 523 nm in PBS with 0.08% sodium azide

fluorescence

λex 554 nm; λem 576 nm in PBS with 0.08% sodium azide

fluorescence

λex 647 nm; λem 669 nm in PBS

fluorescence

-

form

solid

form

lumps

form

-

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

assay

≥80% (sum of isomers, HPCE)

assay

≥80% (sum of isomers, HPCE)

assay

≥80% (HPCE)

assay

≥90% (HPLC), ≥90% (degree of coupling)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. What is the dye:protein ratio used for live cell imaging?

    1 answer
    1. The dye:protein ratio for live cell imaging has not been confirmed. According to literature references, NTA-Atto 488 for live imaging, used for immunofluorescence or staining His-tagged proteins on cells, typically involves dilutions of 1:200 to 1:1000 of the reconstituted dye (1 mg/mL) in buffer.

      Helpful?

  2. Can this reagent detect Histidine affinity tag (HAT)? What is the dilution ratio for WB or IF?

    1 answer
    1. This product is provided as 250 micrograms of powder per vial. The contents of the vial can be reconstituted in 250 microliters of phosphate buffered saline (PBS) to make a 1 mg/mL stock solution. Dilutions of 1:1000 to 1:2500 in PBS with TWEEN 20 (PBS-T; Product P3563 at 1X) can be used to stain histidine-tagged proteins by Western Blot (WB).
      Immunofluorescence (IF) is typically less sensitive than WB, so dilutions of 1:200 to 1:1000 may be appropriate.
      See: https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-labeling-and-modification/ni-nta-atto-conjugates

      Helpful?

  3. Is this product suitable for detection of polyhistidine tagged proteins directly in SDS-PAGE or Western blot?

    1 answer
    1. ATTO 488 NHS-ester can be used for the fluorescence detection of HIS-tagged proteins and polyhistidine tagged proteins directly in SDS-PAGE or Western blot.

      Helpful?

  4. We have a his-tag labeled recombinant protein that we would like to have fluorescently labeled, before we use it to detect/label antibodies. I came across Ni-NTA-Atto Conjugates and I was wondering wether we could use this to label out his-tagprotein?

    1 answer
    1. There should be no difficulty with the use of the product to label a HIS-tagged recombinant protein as long as the HIS-6 tag is in an acceptable position for binding. Typically, this is done with either a N or a C-terminal His6 tag. Please note that the only tests that we conduct on this material include a solubility, purity and a luminescence assay. Hence, we do not offer a detailed protocol for use of the product for HIS-tag protein labeling.

      Helpful?

  5. What is the number of nickel ions present in product 39624? Is it mono-nta, tris-nta, or another form?

    1 answer
    1. There is one nickel ion per molecule, but the exact structure is proprietary.

      Helpful?

  6. What is the molecular weight of this product NTA-ATTO488?

    1 answer
    1. The molecular weight of this product is 964 Da.

      Helpful?

Reviews

No rating value

Active Filters