Skip to Content
Merck
All Photos(1)

Key Documents

40584

Sigma-Aldrich

Amentoflavone

≥98.0% (HPLC)

Synonym(s):

Didemethyl-ginkgetin, I3′,II8-Biapigenin, Tridemethylsciadopitysin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H18O10
CAS Number:
Molecular Weight:
538.46
Beilstein:
380244
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.47

Assay

≥98.0% (HPLC)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)c(c3O2)-c4cc(ccc4O)C5=CC(=O)c6c(O)cc(O)cc6O5

InChI

1S/C30H18O10/c31-15-4-1-13(2-5-15)24-12-23(38)29-21(36)10-20(35)27(30(29)40-24)17-7-14(3-6-18(17)33)25-11-22(37)28-19(34)8-16(32)9-26(28)39-25/h1-12,31-36H

InChI key

YUSWMAULDXZHPY-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Amentoflavone is a naturally occurring polyphenolic biflavonoid found in many plants. Structurally, it has an apigenin dimer which is linked by a C3′-C8′′ covalent bond.

Application

Amentoflavone has been used:
  • as a chemical inhibitor to determine the selective attenuation of p-Cresol glucuronidation in HepaRG cells
  • as a reference standard for qualitative and quantitative analyses of phenolic compounds of Juniperus foetidissima Willd. and Juniperus sabina L. using reverse phase- high-performance liquid chromatography- diode array detector (RP-HPLC-DAD)
  • as a reference standard to analyze and standardize the methanol extract process of Juniperus drupacea Labill. phenolic compounds using reverse phase- high-performance liquid chromatography - diode array detector (RP-HPLC-DAD)

Biochem/physiol Actions

Amentoflavone has various pharmacological properties such as antioxidant, anti-diabetic, anti-tumor, anti-senescence, neuroprotective, and cardioprotective activities.
Biflavonoid with anti-inflammatory, anti-viral and cancer chemopreventive activity. Blocks the induction of COX-2 and up-regulates PPAR-γ. It is a negative modulator of the GABAA receptor at the benzodiazepine binding site.
Biflavonoid with anti-inflammatory, anti-viral and cancer chemopreventive activity. It inhibits vascularization of tumors by blocking the activity of angiogenic VEGFs. Blocks the induction of COX-2 and up-regulates PPAR-γ. It is a negative modulator of the GABAA receptor at the benzodiazepine binding site.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Sasaki et al.
Bioorganic & medicinal chemistry letters, 20(15), 4558-4560 (2010-07-06)
Here, we describe amentoflavone-type biflavonoids, which were isolated from natural sources and were found to inhibit beta-secretase (BACE-1). The structure-activity relationship was studied, and compounds 1-8, 10, 17, and 18 showed BACE-1 inhibitory activity. Among these compounds, 2,3-dihydroamentoflavone 17 and
Sheng Yu et al.
Molecules (Basel, Switzerland), 22(2) (2017-02-18)
Amentoflavone (C30H18O10) is a well-known biflavonoid occurring in many natural plants. This polyphenolic compound has been discovered to have some important bioactivities, including anti-inflammation, anti-oxidation, anti-diabetes, and anti-senescence effects on many important reactions in the cardiovascular and central nervous system
In vitro enzyme inhibitory properties, antioxidant activities, and phytochemical studies on Juniperus drupacea
Orhan DD, et al.,
journal of research in pharmacy, 23(1), 83-92 (2019)
Yuka Kimura et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(1), 429-435 (2009-11-04)
Polyphenols present in foods and supplements may contribute to human health by preventing cardiovascular diseases and cancer. Drug-food or drug-herb interactions have recently come into focus but, except for some phytochemicals, few components of food or herbs participate in such
Janki Prasad et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(14), 1245-1249 (2012-10-23)
In continuation of our drug discovery program on Indian medicinal plants, we isolated bioactive compounds (1-5) from the leaves of Calophyllum inophyllum and evaluated their antidyslipidemic activity in triton induced hyperlipidemia model. The calophyllic acid (1A) and isocalophyllic acid (1B)

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service