Skip to Content
Merck

Skip To

38377

Abberior® STAR 580, NHS ester

for STED application

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥70.0% (degree of coupling)

solubility

DMF: 1 mg/mL, clear

fluorescence

λex 583 nm; λem 605.0 nm±10 nm in PBS, pH 7

storage temp.

−20°C

General description

Abberior STAR 580 is the latest development for STED microscopy with a fluorescent dye in the orange regime. The dye can be excited from 550 to 580 nm. Abberior STAR 580 can substitute dyes like Atto 590/594 or AlexaFluor 584/594. The dye can most effectively be depleted in STED microscopy at 700 to 780 nm via e.g. a Ti:Sa laser or IR diodes.

Abberior STAR 580 is the dye of choice for orange fluorescence. Moreover, the dye is particularly designed and tested for 2-color STED microscopy in combination with Abberior STAR 635 using 2 separate STED wavelength. This offers the great advantage of using 2 regular (not featuring a long Stokes-shift) dyes, which are in general more photostable. Please see our 2-color dye section

Key features
  • Exceptionally bright orange fluorescent dye
  • Ideal for STED microscopy at 700-775 nm
  • 2-color labeling partner with STAR 635P for 2-color STED microscopy


Absorption Maximum, λmax:: 587 nm (PBS, pH 7.4; water)
584 nm (MeOH, aq. ACN)
Extinction Coefficient, ε(λmax):85,000 M-1cm-1 (PBS, pH 7.4)
Correction Factor, CF260 = ε260/εmax: 0.17 (PBS, pH 7.4; water)
Correction Factor, CF280 = ε280/εmax: 0.15 (MeOH, aq. ACN)
Fluorescence Maximum, λfl:607 nm (PBS, pH 7.4)
604 nm (MeOH, aq. ACN)
Recommended STED Wavelength, λSTED:700 - 780 nm
Fluorescence Quantum Yield, η:0.90 (PBS, pH 7.4)
Fluorescence Lifetime, τ: 3.5 ns (PBS, pH 7.4)


Application

Anti-Mouse IgG-Abberior® STAR 580 antibody produced in goat has been used for STED (stimulated emission depletion) microscopy in CA3 (Cornus Ammonis) sections.[1] Abberior® STAR 580 conjugated with secondary antibody has been used for dual-color STED imaging of cultured hippocampal neurons.[2]
Designed and tested for fluorescent super-resolution microscopy

Legal Information

Atto is a trademark of Atto-Tec GmbH
abberior is a registered trademark of Abberior GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
076790468343354
description

for STED application

description

suitable for fluorescence

description

-

description

≥80.0% (degree of coupling)

fluorescence

λex 583 nm; λem 605.0 nm±10 nm in PBS, pH 7

fluorescence

λex 634 nm; λem 654 nm±10 nm in PBS, pH 7.4

fluorescence

λex 638 nm; λem 651 nm±10 nm in H2O

fluorescence

λex 637 nm; λem 660 nm±10 nm in water

solubility

DMF: 1 mg/mL, clear

solubility

DMF: 0.25 mg/mL, clear

solubility

-

solubility

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

assay

≥70.0% (degree of coupling)

assay

-

assay

-

assay

≥80.0% (degree of coupling)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters