Skip to Content
Merck
All Photos(1)

Documents

34440

Sigma-Aldrich

α,α′-Dibromo-p-xylene

purum, ≥98.0% (GC)

Synonym(s):

DMF, 1,4-Bis(bromomethyl)benzene, p-Xylylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Br)2
CAS Number:
Molecular Weight:
263.96
Beilstein:
1100019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

245 °C (lit.)

mp

143-145 °C (lit.)

solubility

dioxane: soluble 1 g/10 mL (hot)

SMILES string

BrCc1ccc(CBr)cc1

InChI

1S/C8H8Br2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H,5-6H2

InChI key

RBZMSGOBSOCYHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α, α′-Dibromo-p-xylene may be used in the preparation of:
  • third-generation bis(1-pyrazolyl)methane ligands
  • 5,5′-[(p-phenyl­enedimethyl­ene)dithio]bis­(1-phen­yl-1H-1,2,3,4-tetra­zole)
  • new bis 1,10-phenanthroline ligand (composed of two subunits of 7-(p-anisyl)-1,10-phenanthroline (phen) joined by p-(CH2)2C6H4(CH2)2 space groups at 4 positions), via Skraup synthesis
  • stereoisomers of α,α′-bis[3-(N,N-diethylcarbamoyl)-piperidino]-p-xylene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jai Young Lee et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 10), o2369-o2370 (2009-01-01)
The title 22-crown-6 unit, C(26)H(28)O(6), comprising of three benzo groups and triethyl-ene glycol, was prepared by the reaction of α,α'-dibromo-p-xylene with 1,8-bis-(2-hydroxy-phen-oxy)-3,6-dioxaoctane in the presence of Cs(2)CO(3) with tetra-hydro-furan (THF) and recrystallized from dichloro-methane-hexane (1:20 v/v) at room temperature. In
D Pomeranc et al.
Journal of the American Chemical Society, 123(49), 12215-12221 (2001-12-06)
A bis-chelating ligand (L1), made of two 7-(p-anisyl)-1,10-phenanthroline (phen) subunits connected with a p-(CH(2))(2)C(6)H(4)(CH(2))(2) spacer through their 4 positions, has been prepared, using Skraup syntheses and reaction of the anion of 4-methyl-7-anisyl-1,10-phenanthroline with alpha,alpha'-dibromo-p-xylene. Its Fe(II) complex, [FeL1(dmbp)](PF(6))(2), was prepared
Daniel L Reger et al.
Inorganic chemistry, 48(3), 936-945 (2009-01-06)
The third-generation bis(1-pyrazolyl)methane ligands p-C(6)H(4)[CH(2)OCH(2)CH(pz)(2)](2) (L(p), pz = pyrazolyl ring) and m-C(6)H(4)[CH(2)OCH(2)CH(pz)(2)](2) (L(m)) have been synthesized by the reaction of (pz)(2)CHCH(2)OH with NaH followed by alpha,alpha'-dibromo-p-xylene or alpha,alpha'-dibromo-m-xylene. The reaction of L(p) with AgBF(4), AgPF(6), and AgO(3)SCF(3) yields the new
5, 5'-[(p-Phenylenedimethylene) dithio] bis (1-phenyl-1H-1, 2, 3, 4-tetrazole).
Wang W, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(4), 1163-1164 (2005)
X Zheng et al.
Chirality, 7(2), 90-95 (1995-01-01)
The stereoisomers of alpha,alpha'-bis[3-(N,N-diethylcarbamoyl)-piperidino]-p-xylene (1) were synthesized. Rac ethyl nipecotate was resolved by diastereomeric (-)-D- and (+)-L-tartrate salt formation. The enantiomeric esters were hydrolyzed to the corresponding nipecotic acids, which were then converted into t-BOC derivatives. Treatment of the latter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service