Skip to Content
Merck

Skip To

17766

DL-α-Glycerol phosphate magnesium salt hydrate

~85% (KT)

Synonym(s):

rac-Glycero-3-phosphate magnesium salt, Magnesium glycerophosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H7MgO6P · xH2O
CAS Number:
Molecular Weight:
194.36 (anhydrous basis)
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
213-149-3
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

~85% (KT)

form

powder

functional group

hydroxyl

lipid type

phosphoglycerides

storage temp.

room temp

SMILES string

O.[Mg++].OCC(O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.Mg.H2O/c4-1-3(5)2-9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;1H2/q;+2;/p-2

InChI key

GLZHYLKPDLVZKJ-UHFFFAOYSA-L

General description

DL-α-Glycerol phosphate is a racemic mixture of the D- and L-glycerophosphate enantiomers.

Application

DL-α-Glycerol phosphate magnesium salt hydrate has been used:
  • in oxidase and dehydrogenase assays
  • as a component of the osteoblast differentiation medium[1]
  • for mineralization of poly(vinyl alcohol) (PVA) hydrogels containing alkaline phosphatase designed for osteochondral regeneration


DL-a-Glycerol phosphate may be used to assess the lysogenic-strain features of agalactiae bacteriophages

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G78866166850020
biological source

synthetic

biological source

-

biological source

-

biological source

-

assay

~85% (KT)

assay

≥93% (GC)

assay

≥95.0% (TLC), 95-102% anhydrous basis (corrected for sodium, T)

assay

≥98.0% (NT)

functional group

hydroxyl

functional group

-

functional group

-

functional group

-

storage temp.

room temp

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

form

powder

form

powder

form

powder or solid

form

solid

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters