Skip to Content
Merck
All Photos(1)

Key Documents

10417

Sigma-Aldrich

8-Anilino-1-naphthalenesulfonic acid ammonium salt

for fluorescence, ≥97.0% (HPLC)

Synonym(s):

1,8-ANS NH4, ANSA, Ammonium 8-anilino-1-naphthalenesulfonate, N-Phenyl peri acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C16H13NO3S · NH3
CAS Number:
Molecular Weight:
316.37
Beilstein:
3581235
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

grade

for fluorescence

Quality Level

Assay

≥97.0% (HPLC)

form

solid

loss

≤2.5% loss on drying

mp

245-253 °C

solubility

H2O: 0.1 g/5mL, clear to very slightly hazy (hot)
NaOH: 1 N
H2O: soluble
acetone: soluble
methanol: soluble

fluorescence

λex 388 nm; λem 470 nm in 0.1 M Tris, 0.2 M KCl, pH 9.0, BSA

SMILES string

N.OS(=O)(=O)c1cccc2cccc(Nc3ccccc3)c12

InChI

1S/C16H13NO3S.H3N/c18-21(19,20)15-11-5-7-12-6-4-10-14(16(12)15)17-13-8-2-1-3-9-13;/h1-11,17H,(H,18,19,20);1H3

InChI key

IPBNQYLKHUNLQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ANS forms an inclusion complex with cyclodextrin. Such model systems are useful to mimic biological recognition and can be studied by measuring the change in fluorescence of free-ANS to complexed-ANS. When ANS enters the hydrophobic core of cyclodestrin, it′s fluorescence increases . Utilized in the reagent phase of a sodium-selective fiber-optic sensor. The reagent phase also contains a copper(II) polyelectrolyte, which binds to ANSA in the absence of sodium and quenches the fluorescence. In the presence of sodium, ANSA forms a cationic complex creating ion-pairs, causing it to fluoresce . ANS is often incorporated into di-block polymers and can be released by changes in the local environment (i.e., temperature, pH, etc.) . ANS is commonly used as a fluorescence probe to investigate molecular assemblies of surfactants and amphiphilic polymers because a blue shift of the emission maximum indicates the fluorophore is located in less polar media . Fluorescent probe for protein studies using methodologies such as steady-state and dynamic fluorescence measurements .
Fluorescent probe for protein studies.
This product is an amphiphilic fluorescent probe for protein studies . Excitation of the unbound dye at 380 nm results in a low fluorescent emission with a maximum at 545 nm. The fluorescence intensity of ANS increases when the dye binds to the hydrophobic regions of a protein . The protein-ANS complex has an emission spectrum which is shifted to a broad maximum at 470 nm. At pH 8, protein causes a 40-fold increase in the relative quantum yield compared to free ANS in solution . ANS has been used to monitor protein conformational changes by binding to the hydrophobic regions of a protein , to gain new insight into protein binding interactions, often by acting as reporter or competitor ligands, to investigate the visual excitation process and structural aspects of photoreceptor cell membranes , and to probe (and disrupt) the structure of both high- and low-density lipoproteins. It has also been used as a substrate in a chemiluminescent enzyme immunoassay system and as a dye for yeast viability determination. The conformational states for apo- and holo- yeast alcohol dehydrogenase were reported under conditions of low pH using ANS fluorescence . ANS is also commonly used as a fluorescence probe to investigate molecular assemblies of surfactants and amphiphilic polymers because a blue shift of its emission maximum indicates the probe is located in less polar environment

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maddalena Collini et al.
Protein science : a publication of the Protein Society, 12(8), 1596-1603 (2003-07-24)
The use of spectroscopy in the study of fatty acids binding to bovine beta-lactoglobulin (BLG) appears to be a difficult task, as these acid compounds, assumed as the protein natural ligands, do not exhibit favorable optical response such as, for
L D'Alfonso et al.
Biochimica et biophysica acta, 1432(2), 194-202 (1999-07-17)
Steady-state and dynamic fluorescence titrations show that: (a) the complex between beta-lactoglobulin (BLG) and 1-anilinonaphthalene-8-sulfonate (ANS) displays a heterogeneous equilibrium with large changes in the binding strength vs. pH and ion concentration; and (b) the fluorescence response of bound ANS
Oktay K Gasymov et al.
Biochemistry, 47(5), 1414-1424 (2008-01-09)
Human tear lipocalin (TL) exhibits diverse functions, most of which are linked to ligand binding. To map the binding site of TL for some amphiphilic ligands, we capitalized on the hydrophobic and hydrophilic properties of 8-anilino-1-naphthalenesulfonic acid (ANS). In single
W P Le et al.
Journal of biochemistry, 119(4), 674-679 (1996-04-01)
Under conditions of low pH, the conformational states of holo-YADH and apo-YADH were examined by protein intrinsic fluorescence, ANS fluorescence, and far-UV CD measurements. The results obtained show that a low ionic strength, with the addition of HCl, the holo-
M Collini et al.
Protein science : a publication of the Protein Society, 9(10), 1968-1974 (2000-12-06)
The fluorescence time decay parameters of the beta-lactoglobulin-1-anilinonaphthalene-8-sulfonate complex have been investigated under physical and chemical perturbations (2 < pH < 8 and added electrolyte 0 < NaCl < 0.5 M) to obtain new insight on the nature of the

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service