Skip to Content
Merck
All Photos(1)

Key Documents

02240

Sigma-Aldrich

Adonitol

BioXtra, ≥99.0% (HPLC)

Synonym(s):

Adonite, Ribitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O5
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720524
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Assay

≥99.0% (HPLC)

form

powder

color

colorless

mp

102-105 °C

solubility

water: 1 g/10 mL, clear to slightly hazy, colorless to very faintly yellow

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

room temp

SMILES string

OC[C@H](O)[C@H](O)[C@H](O)CO
OC[C@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5-

InChI key

HEBKCHPVOIAQTA-ZXFHETKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adonitol (Ribitol), a pentose alcohol, is metabolized to teicholic acids used in the cell walls of gram positive bacteria. Adonitol is often compared to other cell permeating molecules such as formamide, propanediol, and DMSO as a cryopreservation agent.
Adonitol has similar properties like glycerol and is naturally found in Adonis vernalis.

Application

Adonitol has been used as an internal standard for the quantification of polyribosyl ribitol phosphate using high pH anion-exchange chromatography with pulsed amperometric detection (HPAEC/PAD).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General Organic and Biological Chemistry, An integrated approach (2017)
International collaborative study for establishment of the 2nd WHO International Standard for Haemophilus influenzae type b polysaccharide
Fatme M, et al.
Biologicals : Journal of the International Association of Biological Standardization, 43(6), 492-503 (2015)
Freeze Drying (2004)
Anders Østergaard Madsen et al.
The journal of physical chemistry. A, 115(26), 7794-7804 (2011-06-15)
X-ray diffraction data of high quality measured to high resolution on crystals of the two pentitol epimers ribitol (centric) and xylitol (acentric) at 101, 141, and 181 K and data on the two compounds previously recorded at 122 K have
D J Brenner et al.
Journal of clinical microbiology, 15(4), 703-713 (1982-04-01)
DNA relatedness was used to define the biochemical boundaries of Escherichia coli. A large number of biochemically atypical strains were shown to belong to biogroups of E. coli. These included strains negative in reactions for indole, all three decarboxylases, D-mannitol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service