Skip to Content
Merck
All Photos(1)

Documents

01423

Sigma-Aldrich

N-Acetyl-D-penicillamine

for HPLC derivatization, ≥99.0% (T)

Synonym(s):

NAP, NAPA, N-Acetyl-3-mercapto-D-valine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO3S
CAS Number:
Molecular Weight:
191.25
Beilstein:
1724742
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.26

grade

for HPLC derivatization

Assay

≥99.0% (T)

form

powder or crystals

optical activity

[α]20/D +10±2°, c = 1% in ethanol

technique(s)

HPLC: suitable (derivatization)

color

white

mp

185-190 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(=O)N[C@@H](C(O)=O)C(C)(C)S

InChI

1S/C7H13NO3S/c1-4(9)8-5(6(10)11)7(2,3)12/h5,12H,1-3H3,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

MNNBCKASUFBXCO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral reagent for the precolumn derivatization of amino acids or amino alcohols; the diastereoisomers formed can be efficiently resolved by HPLC on conventional reversed-phase columns

Biochem/physiol Actions

N-Acetyl-D-penicillamine (NAP or NAPA), a thiol compound, is often used as a control molecule in studies involving nitric oxide donor molecules such as S-nitroso-N-acetyl-dl-penicillamine (SNAP) and sodium nitroprusside (SNP). N-acetyl-D-penicillamine (NAPA) is also used as a heavy metal chelator.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diederick Duijvesz et al.
PloS one, 8(12), e82589-e82589 (2014-01-07)
Current markers for prostate cancer, such as PSA lack specificity. Therefore, novel biomarkers are needed. Unfortunately, the complexity of body fluids often hampers biomarker discovery. An attractive alternative approach is the isolation of small vesicles, i.e. exosomes, ∼100 nm, which
Chao Qian et al.
Light, science & applications, 9, 59-59 (2020-04-28)
Optical logic operations lie at the heart of optical computing, and they enable many applications such as ultrahigh-speed information processing. However, the reported optical logic gates rely heavily on the precise control of input light signals, including their phase difference
Y Ivashko-Pachima et al.
Molecular psychiatry, 22(9), 1335-1344 (2017-01-25)
Activity-dependent neuroprotective protein (ADNP), vital for brain formation and cognitive function, is mutated in autism and linked to neurodegenerative/psychiatric diseases. An eight-amino-acid peptide snippet of ADNP, NAP (NAPVSIPQ), identified as a smallest active fragment, includes the SxIP microtubule (MT) end-binding
Edward C Jones-López et al.
PLoS medicine, 8(3), e1000427-e1000427 (2011-03-23)
Each year, 10%-20% of patients with tuberculosis (TB) in low- and middle-income countries present with previously treated TB and are empirically started on a World Health Organization (WHO)-recommended standardized retreatment regimen. The effectiveness of this retreatment regimen has not been
Rajan Lamichhane et al.
Virology, 500, 161-168 (2016-11-09)
The TRIM5α protein from rhesus macaques (rhTRIM5α) mediates a potent inhibition of HIV-1 infection via a mechanism that involves the abortive disassembly of the viral core. We have demonstrated that alpha-helical elements within the Linker 2 (L2) region, which lies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service