Skip to Content
Merck
All Photos(1)

Key Documents

39220

Sigma-Aldrich

Dansyl chloride

suitable for fluorescence, BioReagent, ≥99.0% (HPLC)

Synonym(s):

1-Dimethylaminonaphthalene-5-sulfonyl chloride, 5-(Dimethylamino) naphthalene-1-sulfonyl chloride, DNS Chloride, DNSCl, 5-(Dimethylamino)naphthalene-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12ClNO2S
CAS Number:
Molecular Weight:
269.75
Beilstein:
2217205
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Quality Level

Assay

≥99.0% (HPLC)

mp

72-74 °C (lit.)

solubility

acetone: 0.2 g/10 mL

fluorescence

λex 337 nm; λem 492 nm in chloroform (after derivatization with hexylamine)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CN(C)c1cccc2c(cccc12)S(Cl)(=O)=O

InChI

1S/C12H12ClNO2S/c1-14(2)11-7-3-6-10-9(11)5-4-8-12(10)17(13,15)16/h3-8H,1-2H3

InChI key

XPDXVDYUQZHFPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dansyl Chloride, also known as 5-dimethylamino naphthalene-1-sulfonyl chloride, is a pre-column derivatization reagent. [1] After the derivatization reaction, the fluorescent derivatives are analyzed using several analytical methods like NMR, IR, UV-vis, and fluorescence microscopy.[2] Dansyl Chloride is a typical aromatic sulphonyl chloride that reacts with various bases, resulting in derivatives with variable stability.

Application

A fluorogenic reagent for fluorescent N-terminal derivatization of amino acids and peptides and detection by reverse phase HPLC.
Dansyl Chloride is used as a derivatization reagent to study extensive metabolism and low doses of ethinylestradiol administered during pre-clinical studies.[3] Studies conducted to determine bisphenols in human serum through the HPLC method used Dansyl Chloride to obtain the derivative products.[4] Dansyl Chloride derivatives are suitable for LC/MS determination and characterization of glyphosate, aminomethylphosphonic acid (AMPA), and glufosinate from food samples.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martins Jansons et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1177, 122779-122779 (2021-06-08)
Glyphosate and other polar and acidic pesticides have been particularly studied due to the concerns over widespread and intensive use. The chemical properties of these compounds necessitate use of customised methods, such as derivatisation or ion exchange chromatography. These approaches
V Konakovsky et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(4), 408-416 (2011-02-22)
Biogenic amines in wine may impair sensory wine quality and cause adverse health effects in susceptible individuals. In this study, histamine and other biogenic amines were determined by HPLC after amine derivatisation to dansyl chloride conjugates in 100 selected high-quality
Fabio Mazzotti et al.
Journal of mass spectrometry : JMS, 47(7), 932-939 (2012-07-14)
5-Dimethylamino-1-sulfonyl naphthalene (DNS, commonly referred as dansyl) is a functionality, bearing well-established properties in directing the fragmentation, by mass spectrometry (MS), of the corresponding ionized sulfonylated derivatives. This property is shared also by its labeled analogs. The use of d(0)/d(6)
Shaodong Jia et al.
Journal of chromatography. A, 1218(51), 9174-9182 (2011-11-15)
A novel liquid chromatography coupled with quadrupole time-of-flight mass spectrometry (LC-Q-TOFMS) method was developed for the simultaneous determination of 23 amino acids and 7 biogenic amines in food samples. These analytes were pre-column derivatized with dansyl chloride and then separated
Sarah A Goretta et al.
Bioorganic & medicinal chemistry, 20(13), 4012-4019 (2012-06-08)
Sphingomyelin (SM) and cholesterol form microdomains called lipid rafts in cellular membranes. To develop a versatile fluorescent lipid probe, chemical modifications to both the hydrophobic and hydrophilic portions of SM are essential. Few reports describing SM probes with a fluorophore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service