Skip to Content
Merck
All Photos(3)

Key Documents

P4504

Sigma-Aldrich

Potassium chloride

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
powder or crystals

grade

reagent

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

5.5-8.5 (20 °C, 50 g/L)

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium chloride (KCl) is an inorganic salt commonly used in fertilizer industry. Its generation via chlorination of microcline has been reported. The intrinsic and extrinsic conductivity of KCl crystals have been measured. The impact of KCl on the activity of mitochondrial membrane, cristae and matrix proteins has been investigated. It facilitates the preparation of isoxazolines in aqueous medium.

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
Potassium chloride has been used in the preparation of perfusion buffer stock solution (1X).
It has been used in the preparation of the following buffers for small unilamellar vesicles (SUVs) preparation:
  • Escherichia coli supported lipid bilayer (SLB) buffer
  • extract buffer (XB)
  • high-salt extract buffer (HS-XB)

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coculture between hMADS and Mouse Adult CM.
Figeac F, et al.
Bio-protocol, 4(14) (2014)
Using supported bilayers to study the spatiotemporal organization of membrane-bound proteins.
Nguyen PA, et al.
Methods in Cell Biology, 128, 223-241 (2015)
Electrical Conductivity of Potassium Chloride.
Fuller RG, et al.
Physical Review Letters, 20(13), 662-662 (1968)
An environmentally benign synthesis of isoxazolines and isoxazoles mediated by potassium chloride in water.
Han L, et al.
Tetrahedron Letters, 55(14), 2308-2311 (2014)
Suman Mishra et al.
International journal of proteomics, 2015, 647408-647408 (2016-01-20)
Potassium chloride (KCl) has been commonly used in homogenization buffer and procedures of protein extraction. It is known to facilitate release of membrane-associated molecules but the higher concentration of KCl may affect the integrity of mitochondria by breaching the electrostatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service