Skip to Content
Merck
All Photos(4)

Key Documents

F7503

Sigma-Aldrich

Formamide

ReagentPlus®, ≥99.0% (GC)

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.21

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

product line

ReagentPlus®

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 %

technique(s)

hybridization: suitable

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formamide is a polar solvent mainly used for resins and plasticizers. It can be prepared by direct synthesis from carbon monoxide and ammonia.

Application

Formamide can be used as a solvent:
  • To synthesize tetrakis(trialkylsilyl) diphosphate by reacting disodium dihydrogen diphosphate with trialkyl chlorosilane.
  • In Cu/C catalyzed cycloaddition between azides and terminal alkynes to synthesize triazoles.
Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of formamide.
Ladell J and Post B.
Acta Crystallographica, 7(8-9), 559-564 (1954)
The formamide method for the extraction of polysaccharides from haemolytic streptococci.
Fuller AT.
British Journal of Experimental Pathology, 19(2), 130-130 (1938)
Microwave spectrum, structure, dipole moment, and quadrupole coupling constants of formamide.
Kurland RJ and Wilson Jr EB.
J. Chem. Phys. , 27(2), 585-590 (1957)
The dynamics of liquid formamide, N-methylformamide, N,N-dimethylformamide, and N,N-dimethylacetamide. A dielectric relaxation study.
Barthel J, et al.
Journal of Molecular Liquids, 98, 51-69 (2002)
Formamide can dramatically improve the specificity of PCR.
G Sarkar et al.
Nucleic acids research, 18(24), 7465-7465 (1990-12-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service