Skip to Content
Merck
All Photos(1)

Key Documents

F9037

Sigma-Aldrich

Formamide

BioReagent, ≥99.5% (GC), for molecular biology

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.52
grade:
for molecular biology
technique(s):
electrophoresis: suitable
immunohistochemistry: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

product line

BioReagent

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

932 °F

technique(s)

electrophoresis: suitable
immunohistochemistry: suitable

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

suitability

suitable for nucleic acid hybridization

storage temp.

2-8°C

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formamide is a clear liquid amide derived from formic acid. Formamide allows for the denaturation and renaturation of nucleic acids at room temperature.

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.
Formamide is suitable for use in hybridization buffers made using standard recipes. Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Other applications include:

  • Preparation of brain sections of mice for BrdU immunohistochemistry[1]
  • FISH analysis of HeLa cells.[2]
  • Preparation of the polyacrylamide gel for analysis of PCR products using denaturing gradient gel electrophoresis(The Merck Index, 12th ed).

Features and Benefits

  • Deionized for molecular biology applications
  • Packaged under argon gas in amber bottles to protect from oxidation
  • Used for DNA renaturation or DNA-RNA hybridization
  • Reduces thermal stability of double stranded nucleic acids

Suitability

  • nucleic acid hybridization
  • cryoprotectant and gel-stabilizer
  • destabilize nucleic acid duplexes
  • elimination of secondary structure of nucleic acids
  • preparation of hybridization buffers for RNA and DNA application

Other Notes

The product has been deionized for molecular biology use and packaged under argon gas in amber glass bottles to protect it from oxidation.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kaabache et al.
Analytical biochemistry, 232(2), 225-230 (1995-12-10)
The sensitivity of direct solution hybridization of hepatocytes solubilized in guanidium thiocyanate (GuSCN) for detecting alpha 1-acid glycoprotein and albumin mRNAs was studied. The sensitivity of detection was inversely correlated with the DNA concentration. Raising the hybridization temperature from 20
Laurent Geiser et al.
Journal of chromatography. A, 979(1-2), 389-398 (2002-12-25)
A nonaqueous capillary electrophoresis (NACE) method, coupled with either UV or electrospray mass spectrometry (ESI-MS), is described for the simultaneous analysis of seven beta-blockers. The same electrolyte, namely 25 mM ammonium formate and 1 M formic acid, was used with
N C Pagratis
Nucleic acids research, 24(18), 3645-3646 (1996-09-15)
Streptavidin induced electrophoretic mobility shift was used to prepare single stranded (ss) DNA amplified with the polymerase chain reaction in the presence of a biotinylated and a non-biotinylated primer. A variety of denaturing conditions, including incubation at 95 degrees C
Jianwei Li
Journal of pharmaceutical sciences, 91(8), 1873-1879 (2002-07-13)
This article describes the proper selection of extraction solvents to eliminate interference from a polymer matrix to the quantitation of estradiol degradation products in a transdermal formulation by reversed-phase liquid chromatography. The separation is performed by gradient elution with acetonitrile
Helena Mistry et al.
Genome integrity, 1(1), 7-7 (2010-08-04)
DNA double-strand breaks (DSBs) caused by ionizing radiation or by the stalling of DNA replication forks are among the most deleterious forms of DNA damage. The ability of cells to recognize and repair DSBs requires post-translational modifications to histones and

Related Content

Formamide is a reagent that is an ionizing solvent in aqueous buffers. It is widely utilized in biochemistry and molecular biology, particularly in nucleic acids research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service