Skip to Content
Merck
All Photos(1)

Key Documents

95299

Sigma-Aldrich

Hydrogen peroxide solution

purum p.a., ≥35% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

23.3 mmHg ( 30 °C)

Quality Level

grade

purum p.a.

Assay

≥35% (RT)

form

liquid

reaction suitability

reagent type: oxidant

impurities

≤0.02% free acid (as H2SO4)

density

1.13 g/mL at 20 °C

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide is a reactive oxygen species formed as a reaction product of various aerobic metabolic processes. Its ultra-trace quantification in the concentration range of 10-11mole/ml by a fluorometric method has been described.

Application

Hydrogen peroxide solution (dilute alcoholic or aqueous solutions) may be used for the epoxidation of lower olefins in the presence of titanium silicalite (catalyst). It may be used in the synthesis of acenaphthene and peroxyacetic acid.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 3-3 (1994)
Epoxidation of lower olefins with hydrogen peroxide and titanium silicalite.
Clerici MG and Ingallina P.
J. Catal., 140(1), 71-83 (1993)
THE FLUOROMETRIC ANALYSIS OF ULTRAMICRO QUANTITIES OF HYDROGEN PEROXIDE.
A S KESTON et al.
Analytical biochemistry, 11, 1-5 (1965-04-01)
Spectrofluorimetric determination of hydrogen peroxide scavenging activity.
Pazdzioch-Czochra M and Widenska A.
Analytica Chimica Acta, 452(2), 177-184 (2002)
Esther P M Tjin et al.
Cancer immunology research, 2(6), 538-546 (2014-06-05)
In this study, we investigated a large series of immune (escape) markers, relevant to T-cell function, as potential biomarkers for clinical outcome following immunotherapy. We retrospectively studied the expression of immune (escape) markers in metastatic melanoma tissues of 27 patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service