Skip to Content
Merck
All Photos(1)

Documents

95321

Supelco

Hydrogen peroxide solution

≥30%, for trace analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.24

grade

for inorganic trace analysis
for trace analysis

Quality Level

vapor pressure

23.3 mmHg ( 30 °C)

Assay

≥30% (titration)

form

solution

shelf life

~4 yr

concentration

≥30%

impurities

≤2 mg/kg total nitrogen (N)

density

1.11 g/mL at 20 °C

anion traces

bromide (Br-): ≤0.05 mg/kg
chloride (Cl-): ≤0.05 mg/kg
fluoride (F-): ≤1 mg/kg
nitrate (NO3-): ≤0.5 mg/kg
phosphate (PO43-): ≤0.1 mg/kg
sulfate (SO42-): ≤0.1 mg/kg

cation traces

Ag: ≤0.5 μg/kg
Al: ≤1 μg/kg
As: ≤1 μg/kg
Au: ≤1 μg/kg
B: ≤1 μg/kg
Ba: ≤0.5 μg/kg
Be: ≤0.5 μg/kg
Bi: ≤0.5 μg/kg
Ca: ≤10 μg/kg
Cd: ≤0.5 μg/kg
Co: ≤0.5 μg/kg
Cr: ≤0.5 μg/kg
Cs: ≤0.5 μg/kg
Cu: ≤0.5 μg/kg
Fe: ≤5 μg/kg
Ga: ≤0.5 μg/kg
Ge: ≤0.5 μg/kg
Hg: ≤1 μg/kg
In: ≤0.5 μg/kg
Ir: ≤0.5 μg/kg
K: ≤5 μg/kg
Li: ≤0.5 μg/kg
Mg: ≤1 μg/kg
Mn: ≤0.5 μg/kg
Mo: ≤0.5 μg/kg
Na: ≤5 μg/kg
Ni: ≤0.5 μg/kg
Os: ≤0.5 μg/kg
Pb: ≤0.5 μg/kg
Pd: ≤0.5 μg/kg
Pt: ≤0.5 μg/kg
Rb: ≤0.5 μg/kg
Rh: ≤0.5 μg/kg
Ru: ≤0.5 μg/kg
Sb: ≤0.5 μg/kg
Se: ≤1 μg/kg
Sn: ≤0.5 μg/kg
Sr: ≤0.5 μg/kg
Ti: ≤1 μg/kg
Tl: ≤0.5 μg/kg
U: ≤0.5 μg/kg
V: ≤0.5 μg/kg
Zn: ≤1 μg/kg
Zr: ≤0.5 μg/kg

SMILES string

OO

storage temp.

2-8°C

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrogen peroxide solution has been used as a reagent for the chemical digestion of blood, urine, feces and organs from different animals for the analysis of manganese using inductively coupled plasma mass spectrometry (ICP-MS), which serves as a stable endogenous elemental tag to quantify dextran-coated graphene oxide nanoplatelet (GNP-Dex) concentrations in blood, urine, feces and biodistribution studies.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dose ranging, expanded acute toxicity and safety pharmacology studies for intravenously administered functionalized graphene nanoparticle formulations.
Kanakia S, et al.
Biomaterials, 35(25), 7022-7031 (2014)
Koen van der Maaden et al.
Pharmaceutical research, 31(7), 1846-1854 (2014-01-29)
The aim of the study was to develop a cheap and fast method to produce hollow microneedles and an applicator for injecting vaccines into the skin at a pre-defined depth and test the applicability of the system for dermal polio
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
James R Van Brocklyn et al.
Cancer research, 74(19), 5364-5370 (2014-08-12)
Glioblastoma remains a devastating disease for which novel therapies are urgently needed. Here, we report that the Aurora-A kinase inhibitor alisertib exhibits potent efficacy against glioblastoma neurosphere tumor stem-like cells in vitro and in vivo. Many glioblastoma neurosphere cells treated
Osamu Sakai et al.
Investigative ophthalmology & visual science, 56(1), 538-543 (2015-01-13)
The purpose of the present study was to investigate the role of glutathione peroxidase 4 (GPx4) in conjunctival epithelial cells. An immortalized human conjunctival epithelial cell line was used. Cells were transfected with catalase, GPx1, GPx4, SOD1, SOD2, or control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service