Skip to Content
Merck
All Photos(3)

Key Documents

319902

Sigma-Aldrich

Ethyl acetate

Ethyl acetate

ACS reagent, ≥99.5%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥99.5%
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

impurities

H2SO4, passes test (darkened)
≤0.0009 meq/g Titr. acid
≤0.200% water

evapn. residue

≤0.0030%

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate ACS grade solvent meets or exceeds the high standards of the American Chemical Society (ACS). It is ideal for most research needs and delivers replicable, publishable results.
Ethyl acetate ACS grade solvent used in:      
  • Routine chemical synthesis      
  • Drying & purification     
  • Critical labware cleaning

Application

Ethyl acetate (ACS grade) can be used as a solvent in the synthesis of 6-O-acetyl glycosides via selective acetylation of carbohydrates in the presence of a catalytic amount of sulfuric acid. 6-O-acetyl glycoside derivatives are applicable as precursors for the synthesis of complex oligosaccharides.[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A green and regioselective acetylation of thioglycoside with ethyl acetate
Liang Pi-Hui, et al.
Tetrahedron Letters, 51(52), 6928-6931 (2010)
Elif Burcu Bali et al.
Asian Pacific journal of tropical biomedicine, 4(7), 505-514 (2014-09-04)
To investigate the in vitro antimicrobial potential of Thermopsis turcica Kit Tan, Vural & Küçüködük against periodontopathogenic bacteria, its antioxidant activity and cytotoxic effect on various cancer cell lines. In vitro antimicrobial activities of ethanol, methanol, ethyl acetate (EtAc), n-hexane
Chayasith Uttamapinant et al.
Nature protocols, 8(8), 1620-1634 (2013-07-28)
This protocol describes an efficient method to site-specifically label cell-surface or purified proteins with chemical probes in two steps: probe incorporation mediated by enzymes (PRIME) followed by chelation-assisted copper-catalyzed azide-alkyne cycloaddition (CuAAC). In the PRIME step, Escherichia coli lipoic acid
Quinn P Peterson et al.
Nature protocols, 5(2), 294-302 (2010-02-06)
This protocol describes the gram-scale solution-phase synthesis of the colorimetric caspase-3/7 substrate Ac-DEVD-pNA. The caspase enzymes are integral to cellular inflammation and apoptotic cascades, and are commonly studied by cell biologists, medicinal chemists and chemical biologists. In particular, the assessment
Libo Zhou et al.
Journal of visualized experiments : JoVE, (159) (2020-05-26)
A biomimetic NM was developed to serve as a tissue-engineering biological scaffold, which can enhance stem cell anchorage. The biomimetic NM is formed from JBNTs and FN through self-assembly in an aqueous solution. JBNTs measure 200-300 µm in length with

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What methods or procedures should be followed to perform the measurements of 'Residue on Evaporation' and 'Titratable Acid', as outlined in the COA for product 319902?

    1 answer
    1. For an MQ 200 quality level product, it is typically not possible to share test methods. However, in the case of an ACS grade product, one can refer directly to the available ACS monograph.
      The monograph provides specific instructions for conducting the residue on evaporation test, which involves evaporating 40.0 g (45.0 mL) of the sample to dryness in a tared, preconditioned dish on a hot plate (~100 °C). Subsequently, the residue is dried at 105 °C for a duration of 30 minutes.
      Regarding the titratable acid test, the monograph advises adding 0.10 mL of phenolphthalein indicator solution to 10 mL of ethyl alcohol and neutralizing it with 0.01 N sodium hydroxide. Then, 9.0 g (10.0 mL) of the sample should be added, gently mixed, and titrated with 0.01 N sodium hydroxide until the pink color persists for 15 seconds. The requirement is that the usage of sodium hydroxide should not exceed 0.80 mL.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service