1.09623
Ethyl acetate
for analysis EMSURE® ACS,ISO,Reag. Ph Eur
Synonym(s):
Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc
About This Item
for analytical purposes
for extraction
reagent grade
food and beverages
industrial qc
pharmaceutical
sample preparation
Recommended Products
grade
ACS reagent
for analytical purposes
for extraction
reagent grade
Quality Level
Agency
reag. ISO
reag. Ph. Eur.
vapor density
3 (20 °C, vs air)
vapor pressure
73 mmHg ( 20 °C)
product line
EMSURE®
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
801 °F
potency
5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)
expl. lim.
2.2-11.5 %, 38 °F
impurities
≤0.0008 meq/g Titrable acid
≤0.05% Water
≤0.1% Ethanol (GC)
≤0.1% Methanol (GC)
≤0.1% Methyl acetate (GC)
evapn. residue
≤0.001%
color
clearAPHA: ≤10
refractive index
n20/D 1.3720 (lit.)
bp
76.5-77.5 °C (lit.)
mp
−84 °C (lit.)
transition temp
flash point -4 °C
solubility
85.3 g/L
density
0.902 g/mL at 25 °C (lit.)
cation traces
Al: ≤500 ng/g
B: ≤20 ng/g
Ba: ≤100 ng/g
Ca: ≤500 ng/g
Cd: ≤50 ng/g
Co: ≤20 ng/g
Cr: ≤20 ng/g
Cu: ≤20 ng/g
Fe: ≤100 ng/g
Mg: ≤100 ng/g
Mn: ≤20 ng/g
Ni: ≤20 ng/g
Pb: ≤100 ng/g
Sn: ≤100 ng/g
Zn: ≤100 ng/g
SMILES string
CCOC(C)=O
application(s)
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation
format
neat
storage temp.
2-30°C
InChI
1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3
InChI key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Investigating the anti-growth, anti-resistance, and anti-virulence activities of Schoepfia schreberi J.F.Gmel. against the superbug Acinetobacter baumannii.: This study evaluates the effectiveness of plant extracts, possibly involving ethyl acetate as a solvent or extractant, in combating bacterial resistance and virulence, presenting potential applications in analytical methodologies focused on bioactivity assessment (Uc-Cachón et al., 2024).
- Attractant and repellent properties of Senna didymobotrya plant extracts to Amblyomma variegatum and Rhipicephalus appendiculatus.: The research explores natural extract′s efficacy, utilizing ethyl acetate in the extraction process, as eco-friendly pest management solutions, relevant in developing analytical techniques for environmental and agricultural applications (Kinyua et al., 2024).
- Antioxidant activity of Sophora exigua and liposome development of its powerful extract.: This paper discusses the antioxidant properties of plant extracts, with implications for ethyl acetate use in extraction and stabilization processes in pharmaceutical and food industries, enhancing analytical methods for antioxidant capacity measurement (Rodwattanagul et al., 2024).
- In vitro ovicidal and larvicidal activity of a hydroalcoholic extract and its fractions from Cyrtocarpa procera fruits on Haemonchus contortus.: The study investigates ethyl acetate′s role in isolating bioactive compounds with potential applications in the analysis of antiparasitic agents, relevant to both medical and veterinary parasitology (De Jesús-Martínez et al., 2024).
- Antagonistic actions of Paucibacter aquatile B51 and its lasso peptide paucinodin toward cyanobacterial bloom-forming Microcystis aeruginosa PCC7806.: This research focuses on microbial interactions and the role of ethyl acetate in the extraction of peptides, applicable in the analytical examination of microbial ecology and environmental safety (Cha et al., 2024).
Analysis Note
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Titrable acid: ≤ 0.0008 meq/g
Density (d 20 °C/20 °C): 0.901 - 0.904
Boiling point: 76 - 78 °C
Readily carbonizable substances: conforms
Ethanol (GC): ≤ 0.1 %
Methanol (GC): ≤ 0.1 %
Methyl acetate (GC): ≤ 0.1 %
Al (Aluminium): ≤ 500 ng/g
B (Boron): ≤ 20 ng/g
Ba (Barium): ≤ 100 ng/g
Ca (Calcium): ≤ 500 ng/g
Cd (Cadmium): ≤ 50 ng/g
Co (Cobalt): ≤ 20 ng/g
Cr (Chromium): ≤ 20 ng/g
Cu (Copper): ≤ 20 ng/g
Fe (Iron): ≤ 100 ng/g
Mg (Magnesium): ≤ 100 ng/g
Mn (Manganese): ≤ 20 ng/g
Ni (Nickel): ≤ 20 ng/g
Pb (Lead): ≤ 100 ng/g
Sn (Tin): ≤ 100 ng/g
Zn (Zinc): ≤ 100 ng/g
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.05 %
Other Notes
Legal Information
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Learn about our Returnable Solvent Container Program for Western Europe offering solvents in stainless-steel barrels and drums for reduced packaging waste.
Learn about our Returnable Solvent Container Program for Western Europe offering solvents in stainless-steel barrels and drums for reduced packaging waste.
Learn about our Returnable Solvent Container Program for Western Europe offering solvents in stainless-steel barrels and drums for reduced packaging waste.
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service