Skip to Content
Merck
All Photos(1)

Documents

270733

Sigma-Aldrich

Water

suitable for HPLC

Synonym(s):

Aqua, Dihydrogen oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor density

<1 (vs air)

Quality Level

vapor pressure

3 mmHg

form

liquid

technique(s)

HPLC: suitable

evapn. residue

<0.0003%

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

λ

H2O reference

UV absorption

λ: 205 nm Amax: 0.01
λ: 210 nm Amax: 0.01
λ: 250-400 nm Amax: 0.005

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water suitable for HPLC is commonly used as a solvent for isocratic elution in both cation and anion chromatography. Additionally, it is used as a component to prepare standard stock solutions, and for evaporation under a nitrogen stream in a water bath to concentrate the sample before LC-MS-MS analysis.

Application

Water is used as a:
  • Solvent to prepare buffers during the cell lysis process.
  • Component of the mobile phase during the HPLC–UV–diode-array detection for the separation and detection of the analytes.

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Important notice
The article number 270733-6X1L will be discontinued. Please order the single bottle 270733-1L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Identification of the cysteine protease legumain as a potential chronic hypoxia-specific multiple myeloma target gene
Ada-Sophia C et al.
Cells, 11, 292-292 (2022)
Rapid confirmatory assay for the simultaneous detection of ronidazole, metronidazole and dimetridazole in eggs using liquid chromatography-tandem mass spectrometry
Els D et al.
Analyst, 125, 1533-1535 (2000)
Milk ultrafiltrate analysis by ion chromatography and calcium activity for SMUF preparation for different scientific purposes and prediction of its supersaturation
Joseph D et al.
International dairy journal, 68, 60-69 (2017)
Optimization and validation of HPLC-UV-DAD and HPLC-APCI-MS methodologies for the determination of selected PAHs in water samples
Titato, Guilherme T M et al.
Journal of Chromatographic Science, 44, 35-40 (2006)
Manolis Mandalakis et al.
Journal of chromatography. A, 1217(1), 143-150 (2009-12-08)
The analysis of amino acids by gas chromatography mass spectrometry (GC-MS) after their derivatization with N-(t-butyldimethylsilyl)-N-methyltrifluoroacetamide was investigated as an alternative approach for the determination of free (FAA) and combined amino acids (CAA) in aerosols. This technique showed excellent linearity

Articles

Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.

Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.

Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.

Optimized separation of Oligo Standard 6, an internally created HPLC-UV system suitability mix, on Chromolith® RP-18e columns with an evaluation of the effect of flow rates up to 3 mL/min and ion-pairing reagent, TEAA, on the separation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service