Skip to Content
Merck
All Photos(3)

Key Documents

243442

Sigma-Aldrich

Manganese(IV) oxide

ReagentPlus®, ≥99%

Synonym(s):

Manganese dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
form:
crystalline powder
granular
Pricing and availability is not currently available.

grade

reagent

product line

ReagentPlus®

Assay

≥99%

form

crystalline powder
granular

mp

535 °C (dec.) (lit.)

SMILES string

O=[Mn]=O

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(IV) oxide (MnO2) is an eco-friendly chemical having a high theoretical specific capacitance.[1]

Application

Manganese(IV) oxide has been used as a catalyst for the production of gaseous oxygen and liquid water from 30% H2O2 solution.[2]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rigorous buoyancy driven bubble mixing for centrifugal microfluidics.
Burger S, et al.
Lab on a chip, 16(2), 261-268 (2016)
Highly stretchable alkaline batteries based on an embedded conductive fabric.
Abhinav M Gaikwad et al.
Advanced materials (Deerfield Beach, Fla.), 24(37), 5071-5076 (2012-07-05)
Xin Zhao et al.
ACS nano, 6(6), 5404-5412 (2012-05-05)
Manganese dioxide (MnO(2)) particles 2-3 nm in size were deposited onto a porous "activated microwave expanded graphite oxide" (aMEGO) carbon scaffold via a self-controlled redox process. Symmetric electrochemical capacitors were fabricated that yielded a specific capacitance of 256 F/g (volumetric:
Xing Liu et al.
The Analyst, 137(19), 4552-4558 (2012-08-18)
Inorganic nanomaterials that mimic enzymes are fascinating as they potentially have improved properties relative to native enzymes, such as greater resistance to extremes of pH and temperature and lower sensitivity to proteases. Although many artificial enzymes have been investigated, searching
Yongmin He et al.
ACS nano, 7(1), 174-182 (2012-12-20)
A lightweight, flexible, and highly efficient energy management strategy is needed for flexible energy-storage devices to meet a rapidly growing demand. Graphene-based flexible supercapacitors are one of the most promising candidates because of their intriguing features. In this report, we

Questions

1–3 of 3 Questions  
  1. Form says powder/granular. Can you provide an approximate size range for the powder/granular form?

    1 answer
    1. The particle size of this product has not been tested or determined.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service