Skip to Content
Merck
All Photos(2)

Key Documents

377473

Sigma-Aldrich

Manganese(II,III) oxide

97%

Synonym(s):

MnO.Mn2O3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mn3O4
CAS Number:
Molecular Weight:
228.81
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

density

4.8 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mn]O[Mn]O[Mn]=O

InChI

1S/3Mn.4O

InChI key

GVNFAUMGUISVJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II,III) oxide is a transition metal oxide that is formed by annealing manganese oxide in the air above 1000°C. It can be used for a variety of applications such as catalysis, electrochromic devices, and other energy storage applications.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Robinson et al.
Journal of the American Chemical Society, 135(9), 3494-3501 (2013-02-09)
Manganese oxides occur naturally as minerals in at least 30 different crystal structures, providing a rigorous test system to explore the significance of atomic positions on the catalytic efficiency of water oxidation. In this study, we chose to systematically compare
Chemistry of elements (2012)
Lucas K Wagner
The Journal of chemical physics, 138(9), 094106-094106 (2013-03-15)
Very accurate wave functions are calculated for small transition metal oxide molecules. These wave functions are decomposed using reduced density matrices to study the underlying correlation of electrons. The correlation is primarily of left-right type between the transition metals and
Andres M Cardenas-Valencia et al.
Rapid communications in mass spectrometry : RCM, 27(5), 635-642 (2013-02-16)
In situ analytical techniques that require the storage and delivery of reagents (e.g., acidic or basic solutions) have inherent durability limitations. The reagentless electrolytic technique for pH modification presented here was developed primarily to ease and to extend the longevity
Y Zhang et al.
Journal of hazardous materials, 248-249, 81-88 (2013-01-23)
Several MCM-41 materials were synthesized at different conditions by hydrothermal procedure using cheap and easily available industrial water glass as silica source. Fe doped manganese-based oxide/MCM-41 sorbents were prepared by a sol-gel method. The effects of loadings of metal oxide

Articles

Synthesis, Properties, and Applications of Perovskite-Phase Metal Oxide Nanostructures

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service