Skip to Content
Merck
All Photos(3)

Key Documents

220736

Sigma-Aldrich

Florisil®

100-200 mesh

Synonym(s):

Activated magnesium silicate, Magnesium silicate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
SB.52

form

powder

Quality Level

technique(s)

gas chromatography (GC): suitable

particle size

100-200 mesh

transition temp

transition temp 650 °C (activation temperature)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An activated magnesium silicate used as an adsorbent in gas chromatography.

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Gefen et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 182, 58-63 (2014-12-17)
Scorpions exhibit some of the lowest recorded water loss rates among terrestrial arthropods. Evaporative water loss to the surrounding environment occurs mainly through the integument, and thus its resistance to water loss has paramount significance for the ability of scorpions
Separation of lipid classes by chromatography on florisil.
Carroll KK.
Journal of Lipid Research, 2(2), 135-141 (1961)
Alain Hildebrandt et al.
Archives of environmental contamination and toxicology, 57(2), 247-255 (2008-12-05)
This study was aimed to assess the presence and fate of 22 organochlorinated pesticides (OCHs) and their degradation products and 16 Environmental Protection Agency-priority polycyclic aromatic hydrocarbons (PAHs) in soils of the Ebro River basin (NE Spain) during a 3-year
Youtao Xie et al.
Acta biomaterialia, 5(6), 2331-2337 (2009-04-14)
In this paper, chemically synthesized Mg(2)SiO(4) (MS) powder was plasma-sprayed onto a titanium alloy substrate to evaluate its application potentials in biomedicine. The phase composition and surface morphology of the MS coating were analyzed. Results showed that the MS coating
U Avsar et al.
Cellular and molecular biology (Noisy-le-Grand, France), 59 Suppl, OL1835-OL1841 (2013-02-13)
This study evaluated the protective effect of Panax Ginseng (PG) on bone metabolism in an experimental ovariectomy (OVX) model of osteoporosis in which inflammation was induced by subcutaneous magnesium silicate. The groups were: sham control (Group1, SH), sham+inflammation (Group2, SHinf)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service