Skip to Content
Merck
All Photos(4)

Key Documents

217565

Sigma-Aldrich

Barium chloride dihydrate

ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder, crystals or chunks

grade

ACS reagent

Assay

≥99%

form

powder, crystals or chunks

impurities

≤0.005% insolubles
≤0.005% oxidizers

loss

14.0-16.0% loss on drying

pH

5.2-8.2 (25 °C, 5%)

cation traces

Ca: ≤0.05%
Fe: ≤2 ppm
K: ≤0.0025%
Na: ≤0.005%
Sr: ≤0.1%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Barium chloride dihydrate has been used:
  • in patch-clamp electrophysiological studies to block subthreshold-activated potassium channels
  • as an inward rectifier potassium (Kir) channel blocker to study the role of the channel by measuring the microglial membrane currents in response to voltage ramp stimuli after ATP injection
  • to prepare the stock solution to make artificial cerebrospinal fluid (aCSF)

Biochem/physiol Actions

Barium chloride is a potassium ion channel blocker. It is considered toxic. Barium chloride prevents the D2-like receptor-mediated hyperpolarisation of spinal central pattern generator (CPG) neurons

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence D Picton et al.
Scientific reports, 6, 35749-35749 (2016-10-21)
Dopamine plays important roles in the development and modulation of motor control circuits. Here we show that dopamine exerts potent effects on the central pattern generator circuit controlling locomotory swimming in post-embryonic Xenopus tadpoles. Dopamine (0.5-100 μM) reduced fictive swim bout
Evaluating the performance of peroxide and conjugated diene values in monitoring quality of used frying oils.
Farhoosh R and Moosavi SMR.
Journal of Agriculture, Science and Technology, 11, 173-179 (2010)
Proton magnetic resonance study of the structure of barium chloride dihydrate.
Silvidi AA and McGrath JW.
J. Chem. Phys. , 32(3), 924-926 (1960)
Bokyung Song et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Cytokines are important neuroinflammatory modulators in neurodegenerative brain disorders including traumatic brain injury (TBI) and stroke. However, their temporal effects on the physiological properties of microglia and neurons during the recovery period have been unclear. Here, using an ATP-induced cortical
Evaluation of quality changes in walnut kernels (Juglans regia L.) by Vis/NIR spectroscopy.
Jensen PN, et al.
Journal of Agricultural and Food Chemistry, 49(12), 5790-5796 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service