Skip to Content
Merck
All Photos(2)

Documents

529591

Sigma-Aldrich

Barium chloride dihydrate

≥99.999% trace metals basis

Synonym(s):

Barium(2+) dichloride dihydrate, Dichlorobarium dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.999% trace metals basis

form

powder

impurities

≤10.0 ppm Trace Metal Analysis

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Growth and characterization of barium chloride dihydrate crystal: This study explores the coordination of barium chloride dihydrate with amino acids, highlighting its applications in nonlinear optical crystalline compounds (E Peter et al., 2018).
  • Growth and characterization of a semiorganic NLO material: Describes the growth of L-histidine barium chloride dihydrate crystals by slow solvent evaporation, emphasizing its potential in non-linear optical applications (TR Beena et al., 2016).
  • Material Synthesis, Energy Diagram and Physicochemical Properties of Frequency Doubling 2-Aminopyridine Barium Chloride (2APBC) Crystal: Examines the synthesis and optical properties of 2-aminopyridine barium chloride crystals, useful for frequency doubling applications (D Sivavishnu et al., 2019).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Junsung Woo et al.
Experimental neurobiology, 28(1), 30-42 (2019-03-12)
The neuronal activity-dependent change in the manner in which light is absorbed or scattered in brain tissue is called the intrinsic optical signal (IOS), and provides label-free, minimally invasive, and high spatial (~100 µm) resolution imaging for visualizing neuronal activity
Fan Hu et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(12), 1051-1058 (2012-08-15)
Danshen (Salviae Miltiorrhizae Radix) and Gegen (Puerariae Lobatae Radix) have been widely used in treating cardiovascular diseases for thousands of years in China. The present study was carried out to evaluate the effects of a Danshen and Gegen decoction (DG)
Laura A Merriam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4614-4622 (2013-03-08)
After G-protein-coupled receptor activation and signaling at the plasma membrane, the receptor complex is often rapidly internalized via endocytic vesicles for trafficking into various intracellular compartments and pathways. The formation of signaling endosomes is recognized as a mechanism that produces
Virginia Bay et al.
Glia, 60(4), 651-660 (2012-02-01)
Uptake of K(+) released by axons during action potential propagation is a major function of astrocytes. Here, we demonstrate the importance of glial inward rectifying potassium channels (Kir) in regulating extracellular K(+) ([K(+)](o)) and axonal electrical activity in CNS white
Hanna Heikkinen et al.
Investigative ophthalmology & visual science, 53(9), 5653-5664 (2012-06-30)
To examine the amplification and kinetics of murine rod photoresponses by recording ERG flash responses in vivo and ex vivo from the same retina. We also aimed to evaluate the two available methods for isolating the rod signal from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service