Skip to Content
Merck
All Photos(4)

Documents

208043

Sigma-Aldrich

Sodium sulfide nonahydrate

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S·9H2O
CAS Number:
Molecular Weight:
240.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

crystals or chunks

reaction suitability

reagent type: catalyst
core: sodium

color

colorless to slightly yellow

anion traces

SO32- and S2O32-: ≤0.1%

cation traces

Fe:, passes test
NH4+: ≤0.005%

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.O.O.[Na]S[Na]

InChI

1S/2Na.9H2O.S/h;;9*1H2;

InChI key

UIINQVYGFKNSPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfide nonahydrate participates in the conversion of nitroxides to amines, via reduction.

Application

Sodium sulfide nonahydrate has been used as a source of H2S, to study the effect of H2S on amyloid fibrils associated with neurodegenerative diseases. It has been used in the preparation of sulfide standards for the quantitative analysis of sulfides in raw wastewater (sewage) samples.
It serves as a catalyst during synthesis of thioamides.
Sodium sulfide nonahydrate has been used as a standard for the quantification of volatile sulfur compounds in cheese samples by gas chromatography. It reacts with 4-(2′-hydroxyaryl)-2-(N,N-dialkylamino)-1,3-dithiolium perchlorates in ethanol at room temperature and boiling ethanol to form 1,3-dithiole-2-thiones and 2′-hydroxyacetophenones, respectively. Sodium sulfide nonahydrate may be used as a reducing agent for the conversion of vic-dibromides to the corresponding olefins via reductive debromination.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preservation and simultaneous analysis of relevant soluble sulfur species in sewage samples
Keller-Lehmann B, et al.
Proceedings of the 2nd International Conference on Intelligent Technologies and Engineering Systems (ICITES2013). Springer International Publishing, 2014., 26 (2006)
Daniel L Priebbenow et al.
Chemical Society reviews, 42(19), 7870-7880 (2013-06-25)
The Willgerodt-Kindler reaction has, in recent years, received limited attention and application in synthetic organic chemistry. With the advent of new technology such as microwave-assisted heating, several new, high-yielding, practical, and more environmentally friendly reaction protocols have been developed. This
Reaction of 4-(2'-Hydroxyaryl)-1, 3-dithiolium Salts with Sodium Sulfide. A Selective Synthesis of 2'-Hydroxyacetophenones.
Birsa ML.
Synthetic Communications, 33(17), 3071-3076 (2003)
A new reduction system: combination of sodium sulfide (sodium hydrosulfide) with phase transfer agent in a two-phase mixture. Reductive debromination ofvic-dibromides to olefins.
Nakayama J, et al.
Tetrahedron Letters, 24(29), 3001-3004 (1983)
Reduction of nitroxides to amines by sodium sulfide.
Kornblum N and Pinnick HW.
The Journal of Organic Chemistry, 37(12), 2050-2051 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service