Skip to Content
Merck
All Photos(4)

Key Documents

203734

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
99.99% trace metals basis
form:
crystals
Pricing and availability is not currently available.

Quality Level

Assay

99.99% trace metals basis

form

crystals

pH

3.5-6 (25 °C, 50 g/L)

mp

58 °C (lit.)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) chloride tetrahydrate is a hydrated metal chloride.3 Its toxicity in Swiss mice has been assessed.[1] It participates in the preparation of metal(II) complexes of mixed ligands-dimethydithiocarbamic and p-aminobenzoic acids.3

Application

Manganese(II) chloride tetrahydrate has been used in a loop-mediated isothermal amplification (LAMP) method for the detection of nucleic acids.[2]
Manganese(II) chloride tetrahydrate may be used:
  • As a promoter for the microwave-assisted synthesis of dihydropyrimidin-2(1H)-ones.[3]
  • To synthesize a manganese(II) adipate coordinate complex with antiferromangentic property below 15K.[4]
  • To synthesize active manganese oxide with potential application as an oxidant for organic compounds.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna Lucio et al.
Frontiers in aging neuroscience, 11, 331-331 (2019-12-24)
Parkinson's disease (PD) is a neurodegenerative disease with a complex etiology. Several factors are known to contribute to the disease onset and its progression. However, the complete underlying mechanisms are still escaping our understanding. To evaluate possible correlations between metabolites
Hydrothermal synthesis and magnetic behavior of a novel layered coordination polymer generated from manganese (II) adipate.
Kim YJand Jung DY
Inorganic Chemistry, 39(7), 1470-1475 (2000)
Active manganese dioxide on silica: Oxidation of alcohols under solvent-free conditions using microwaves.
Varma RS, et al.
Tetrahedron Letters, 38(45), 7823-7824 (1997)
Yi Zhang et al.
Analytical chemistry, 86(14), 7057-7062 (2014-06-18)
This report demonstrates a straightforward, robust, multiplexed and point-of-care microcapillary-based loop-mediated isothermal amplification (cLAMP) for assaying nucleic acids. This assay integrates capillaries (glass or plastic) to introduce and house sample/reagents, segments of water droplets to prevent contamination, pocket warmers to
Cobalt (II) chloride or manganese (II) chloride or tin (II) chloride promoted one pot synthesis of dihydropyrimidin-2 (1H)-ones using microwave irradiation.
Kumar S, et al.
Indian Journal of Chemistry, 44B, 762-767 (2005)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Hello! In COA for Manganese(II) chloride tetrahydrate cat 203734 we see only Quality Release Date. Could you please confirm that is expire date for this reagent?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service