Skip to Content
Merck
All Photos(1)

Key Documents

300410

Millipore

Digitonin, High Purity

Digitonin, High Purity

CAS 11024-24-1 is a non-ionic detergent commonly used to solubilize membrane-bound proteins.

Synonym(s):

Digitonin, High Purity, Digitin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H92O29
CAS Number:
Molecular Weight:
1229.31
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 3161

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

aggregation number

60 - 70

color

white

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O[C@@H]%11OC[C@H]([C@@H]([C@H]%11O)O)O)O[C@@H]9O[C@@H]([C@@H]([C@@H]([C@H]9O)O[C@@H]%10O[C@@H]([C@H]([C@@H]([C@H]%10O)O)O)CO)O)CO)O[C@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@H]3[C@@H](C[C@@]4([C@@H]5[C@H]([C@H]6[C@@]([C

InChI

1S/C56H92O29/c1-19-7-10-56(75-17-19)20(2)31-45(85-56)37(67)32-22-6-5-21-11-26(24(61)12-55(21,4)23(22)8-9-54(31,32)3)76-50-42(72)39(69)44(30(16-60)80-50)81-53-48(47(36(66)29(15-59)79-53)83-49-40(70)33(63)25(62)18-74-49)84-52-43(73)46(35(65)28(14-58)78-52)82-51-41(71)38(68)34(64)27(13-57)77-51/h19-53,57-73H,5-18H2,1-4H3/t19?,20-,21-,22+,23-,24+,25+,26+,27+,28+,29+,30+,31-,32+,33-,34+,35-,36+,37-,38-,39+,40+,41+,42+,43+,44-,45+,46-,47-,48+,49-,50+,51-,52-,53-,54+,55-,56+/m0/s1

InChI key

UVYVLBIGDKGWPX-QYLWGQLPSA-N

General description

A non-ionic detergent commonly used to solubilize membrane-bound proteins.
Non-ionic detergent commonly used to solubilize membrane-bound proteins. Aggregation number: 60-70.

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.
Reconstitute at 1 g/20 ml in boiling H₂O. Heat for 15 min at 98°C. Cool and place in the refrigerator (4°C) for 3 h. Filter off any precipitate.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naotaka Tsutsumi et al.
eLife, 9 (2020-08-09)
Frizzleds (Fzd) are the primary receptors for Wnt morphogens, which are essential regulators of stem cell biology, yet the structural basis of Wnt signaling through Fzd remains poorly understood. Here we report the structure of an unliganded human Fzd5 determined
Tsung-Han Chou et al.
Cell, 182(2), 357-371 (2020-07-02)
Excitatory neurotransmission meditated by glutamate receptors including N-methyl-D-aspartate receptors (NMDARs) is pivotal to brain development and function. NMDARs are heterotetramers composed of GluN1 and GluN2 subunits, which bind glycine and glutamate, respectively, to activate their ion channels. Despite importance in
Herlon Victor Rodrigues Silva et al.
Biopreservation and biobanking, 18(5), 415-424 (2020-08-12)
Anhydrous preservation is a promising approach for storage of living biomaterials at nonfreezing temperatures. Using the domestic cat model, the objectives of this study were to characterize changes in histology, DNA integrity, and viability of testicular tissues from adult versus
Judith Miné-Hattab et al.
eLife, 10 (2021-02-06)
In response to double strand breaks (DSB), repair proteins accumulate at damaged sites, forming membrane-less sub-compartments or foci. Here we explored the physical nature of these foci, using single molecule microscopy in living cells. Rad52, the functional homolog of BRCA2
Maria Maldonado et al.
eLife, 9 (2020-08-26)
Respiration, an essential metabolic process, provides cells with chemical energy. In eukaryotes, respiration occurs via the mitochondrial electron transport chain (mETC) composed of several large membrane-protein complexes. Complex I (CI) is the main entry point for electrons into the mETC.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service