Skip to Content
Merck
All Photos(1)

Documents

17093

Sigma-Aldrich

Ammonium hydroxide solution

puriss. p.a. plus, ≥25% NH3 in H2O

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060103
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a. plus

Quality Level

form

liquid

expl. lim.

27 %

concentration

≥25% NH3 in H2O

evapn. residue

≤0.001%

pH

11.7 (20 °C)

density

0.91 g/mL at 20 °C
0.9 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.2 mg/kg
phosphate (PO43-): ≤0.2 mg/kg
sulfate (SO42-): ≤1 mg/kg

cation traces

Ag: ≤0.01 mg/kg
As, Sb (as As): ≤0.01 mg/kg
Au: ≤0.01 mg/kg
B: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤0.01 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.01 mg/kg
Cs: ≤0.01 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.01 mg/kg
Ga: ≤0.01 mg/kg
Ge: ≤0.01 mg/kg
In: ≤0.01 mg/kg
K: ≤0.01 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.01 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.01 mg/kg
Na: ≤0.01 mg/kg
Ni: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pt: ≤0.01 mg/kg
Sn: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Ti: ≤0.01 mg/kg
Tl: ≤0.01 mg/kg
V: ≤0.01 mg/kg
Zn: ≤0.01 mg/kg
Zr: ≤0.01 mg/kg

SMILES string

[NH4+].[OH-]

storage temp.

2-8°C

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium hydroxide is a weak base mainly used as a source to prepare other ammonium compounds like urea, ammonium carbonate and ammonium sulfate etc.
Ammonium hydroxide solution is a solution of 30-33% NH3 in H2O. It is a colorless liquid having a strong characteristic odor. Electrolytic deposition of silver onto vitreous carbon electrodes from ammonium hydroxide solutions by the potential step technique has been reported.

Application

Ammonium hydroxide solution may be used in the following studies:
  • Preparation of magnetic nanoparticles.
  • To investigate the effect of substrate on the morphology of ZnS thin films by chemical bath deposition.
  • Preparation of amine functionalized cellulose nanocrystals (CNC-NH2).
  • Wet processing of the GaAs(100) surface.
Ammonium hydroxide solution may be used:
  • As a medium to prepare magnetite (Fe3O4) nanoparticles via a modified controlled chemical coprecipitation method.
  • As a solvent and reagent to prepare 4-aryl and 4-alkyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylates via Hantzsch synthesis in a microwave cavity synthesizer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient and fast procedure for the Hantzsch dihydropyridine synthesis under microwave conditions.
Oehberg L and Westman J
Synlett, 2001(08), 1296-1298 (2001)
Ramakrishna A
Goyal?s IIT foundation course chemistry: For Class-9, 160-160 (2014)
Synthesis and characterization of biocompatible Fe3O4 nanoparticles.
Sun J, et al.
Journal of Biomedical Materials Research Part A, 80(2), 333-341 (2007)
Claudia M Gómez-Gutiérrez et al.
Scanning, 37(6), 389-392 (2015-05-27)
In this work, the influence of substrate on the morphology of ZnS thin films by chemical bath deposition is studied. The materials used were zinc acetate, tri-sodium citrate, thiourea, and ammonium hydroxide/ammonium chloride solution. The growth of ZnS thin films
Synchrotron photoemission spectroscopy study of ammonium hydroxide etching to prepare well-ordered GaAs (100) surfaces.
Lebedev MV, et al.
Applied Surface Science, 229(1), 226-232 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service