Skip to Content
Merck
All Photos(2)

Key Documents

16211

Sigma-Aldrich

Tetrachloroethylene

≥99.5%

Synonym(s):

PCE, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Beilstein:
1361721
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
Assay:
≥99.5%
technique(s):
NMR: suitable
bp:
121 °C (lit.)
vapor pressure:
13 mmHg ( 20 °C)
19 mmHg ( 25 °C)
Pricing and availability is not currently available.

vapor density

5.83 (vs air)

Quality Level

vapor pressure

13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

Assay

≥99.5%

form

liquid

contains

stabilizer

technique(s)

NMR: suitable

impurities

≤0.001% free acid (as HCl)
≤0.001% non-volatile matter
≤0.01% water (Karl Fischer)

refractive index

n20/D 1.505 (lit.)

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

SMILES string

Cl\C(Cl)=C(\Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrachloroethylene is a chlorinated ethene derivative mainly found in the contaminated groundwater[1] due to its extensive use as a dry cleaning solvent.[2] Its sorption and reduction kinetics with zero-valent iron has been determined in a closed, anaerobic batch system.[3]
Tetrachloroethylene, a chlorinated ethylene, is a common industrial solvent. It is also one of the major contaminant found in groundwater supplies.[4]

Application

TCE may be used as a metal cleaning or degreasing agent.[5]
Tetrachloroethylene (TCE) was employed as solvent to investigate the NMR spectra of end functionalized cyclopentadienyl poly(ethylene) (PE-Cp).[6]

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Cyclopentadienyl Capped Polyethylene and Subsequent Block Copolymer Formation Via Hetero Diels-Alder (HDA) Chemistry.
Espinosa E, et al.
Macromolecular Rapid Communications, 32(18), 1447-1453 (2011)
Use of exhaled air as an improved biomonitoring method to assess perchloroethylene short-term exposure.
Dias CM, et al.
Environmental Research, 156, 108-112 (2017)
Dechlorination of tetrachloroethylene by palladized iron in the presence of humic acid.
Doong RA and Lai TJ.
Water Research, 39(11), 2309-2318 (2005)
Source apportionment of groundwater pollution in a city's eastern part using multivariate statistical techniques.
Jiang P, et al.
IOP Conference Series: Earth and Environmental Science., 59(1) (2017)
Sorption of trichloroethylene and tetrachloroethylene in a batch reactive metallic iron-water system.
D R Burris et al.
Environmental science & technology, 29(11), 2850-2855 (1995-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service