Skip to Content
Merck
All Photos(2)

Documents

12322

Sigma-Aldrich

Iron(III) chloride solution

greener alternative

purum, 45% FeCl3 basis

Synonym(s):

Ferric chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Quality Level

grade

purum

form

liquid

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

45% (FeCl3)

greener alternative category

SMILES string

Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) chloride is a mild oxidizing agent. It is widely employed as a Lewis acid in organic synthesis. On crystallization with water, it forms hydrates. It can be prepared by passing chlorine over scrap iron at approximately 650°C.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. The iron catalysts are known to be inexpensive and environmentally benign. Click here for more information.

Application

Iron(III) chloride may be used for the polymerization of pyrrole and for the preparation of the phenanthrene ring. It is widely employed for the following reactions:
  • polymerizations
  • oxidations
  • oxidative couplings
  • reductions
  • C−C bond formation
  • Ferrier rearrangement
  • one-pot multicomponent condensations
  • Friedel-Crafts reactions
  • cyclization′s
  • glycosidation
  • Prins-type cyclisation

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimum reaction conditions for the polymerization of pyrrole by iron (III) chloride in aqueous solution.
Armes SP.
Synthetic Metals, 20(3), 365-371 (1987)
Recent uses of iron (III) chloride in organic synthesis.
Diaz DD, et al.
Current Organic Chemistry, 10(4), 457-476 (2006)
Kailiang Wang et al.
The Journal of organic chemistry, 74(2), 935-938 (2008-12-05)
Easily available and nontoxic FeCl(3) catalyzes intramolecular oxidative coupling for the direct construction of the phenanthrene ring using meta-chloroperbenzoic acid as sole oxidant at room temperature in excellent yields. The mechanistic investigations show that FeCl(3)-catalyzed coupling proceeds through the heterolytic
Eagleson M.
Concise Encyclopedia Chemistry, 553-553 (1994)
Abdelwareth A O Sarhan et al.
Chemical Society reviews, 38(9), 2730-2744 (2009-08-20)
In this critical review, the use of iron(III) chloride in oxidative C-C couplings of arenes and related unsaturated compounds is presented and reviewed. The approach allows highly selective dimerisations of phenol derivatives, naphthols, and heterocyclic compounds. Sequential couplings give access

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service