Skip to Content
Merck
All Photos(1)

Key Documents

126799

Sigma-Aldrich

1-Dodecanol

reagent grade, 98%

Synonym(s):

Alcohol C12, Dodecyl alcohol, Lauryl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
Beilstein:
1738860
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39020229
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
bp:
260-262 °C (lit.)
vapor pressure:
0.1 mmHg ( 20 °C)

grade

reagent grade

Quality Level

vapor density

7.4 (vs air)

vapor pressure

0.1 mmHg ( 20 °C)

Assay

98%

form

solid or liquid

autoignition temp.

500 °F

expl. lim.

4 %

refractive index

n20/D 1.442 (lit.)

bp

260-262 °C (lit.)

mp

22-26 °C (lit.)

solubility

water: slightly soluble 1 g/L at 23 °C

density

0.833 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCO

InChI

1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3

InChI key

LQZZUXJYWNFBMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Dodecanol, a fatty alcohol, is a long chain aliphatic alcohol. Solid-liquid phase diagrams of its binary mixtures with various other fatty alcohols (1-tetradecanol, 1-hexadecanol and 1-octadecanol) have been evaluated by employing high pressure microscopy and DSC (Differential scanning calorimetry). It undergoes oxidation in the presence of multiwall carbon nanotubes (MWCNTs) supported MoO3 (Mo/MWCNTs) and potassium containing Mo/MWCNTs (K/Mo/MWCNTs) catalysts to afford 1-dodecanal. Its oxidation in the presence of Ru and Pt based carbon catalysts has been reported.

Application

1-Dodecanol may be employed as an extraction reagent in the pipette vial dispersive liquid-liquid microextraction method coupled with HPCL studies.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium-promoted Mo/MWCNTs catalysts for effective partial oxidation of 1-dodecanol to 1-dodecanal.
Liao SY and Ko AN.
Applied Catalysis A: General, 496, 79-85 (2015)
Xuefei Xi et al.
Journal of separation science, 39(2), 391-398 (2015-11-04)
A simple, sensitive, and efficient method of using a pipette vial to perform dispersive liquid-liquid microextraction based on the solidification of floating organic droplets was coupled with high-performance liquid chromatography (HPLC) and a diode array detector for the preconcentration and
High Pressure Solid-Liquid Equilibrium of Fatty Alcohols Binary Systems from 1-Dodecanol, 1-Tetradecanol, 1-Hexadecanol, and 1-Octadecanol.
Dorighello Carareto ND, et al.
Journal of Chemical and Engineering Data, 60(10), 2966-2973 (2015)
Fragrances by selective oxidation of long-chain alcohols.
Villa A, et al.
Chinese Journal of Catalysis, 35(6), 945-951 (2014)
Chris Grant et al.
Biotechnology and bioengineering, 109(9), 2179-2189 (2012-04-05)
This article describes the first reported microwell whole-cell bioconversion using a water immiscible substrate that matches the specific activity and yield achieved in a 1.2 L stirred tank bioreactor. Maximum yields of 0.6 g/L(total) 1-dodecanol achieved in 24 h compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service