Skip to Content
Merck
All Photos(1)

Documents

11213

Sigma-Aldrich

Ammonium bicarbonate

puriss., meets analytical specification of Ph.Eur., BP, E 503, 99-101%

Synonym(s):

Ammonium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HCO3
CAS Number:
Molecular Weight:
79.06
Beilstein:
4329606
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38020216
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.7 (vs air)

vapor pressure

513 hPa ( 50 °C)
67.1 hPa ( 20 °C)

grade

puriss.

Assay

99-101%

form

powder or crystals

quality

meets analytical specification of Ph.Eur., BP, E 503

impurities

residual solvents, complies
tar like substances, complies
≤0.001% heavy metals (as Pb)
≤0.01% non-volatile matter

pH

7-8.5 (25 °C, 79.1 g/L)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤30 mg/kg
sulfide (S2-): in accordance

cation traces

As: ≤1 mg/kg
Fe: ≤5 mg/kg
Hg: ≤1 mg/kg
Pb: ≤5 mg/kg

storage temp.

room temp

SMILES string

N.OC(O)=O

InChI

1S/CH2O3.H3N/c2-1(3)4;/h(H2,2,3,4);1H3

InChI key

ATRRKUHOCOJYRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium bicarbonate is bicarbonate salt of the ammonium ion. In addition to being a source of ammonia, it is also commonly used as a mild base. Bicarbonate of ammonia appears as granular crystals and is easily soluble in water. In the synthesis of catalysts, ammonium bicarbonate is used.

Application

Ammonium bicarbonate is used:
  • As a mineral fuel to prepare alumina-zirconia nano powders by sol-gel combustion.
  • Precipitating agent in the synthesis of γ-Al2O3 sample (γ-Al2O3-1), that is used for the removal of fluoride from aqueous solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Su et al.
ChemSusChem, 8(5), 813-816 (2015-02-11)
A highly efficient, reversible hydrogen storage-evolution process has been developed based on the ammonium bicarbonate/formate redox equilibrium over the same carbon-supported palladium nanocatalyst. This heterogeneously catalyzed hydrogen storage system is comparable to the counterpart homogeneous systems and has shown fast
Kaina Zhang et al.
Mikrochimica acta, 186(2), 68-68 (2019-01-11)
A novel magnetic nanomaterial for use in metal ion based affinity chromatography is described. It is based on the chelation between the phosphate groups of phytic acid (PA) and Ti(IV) ions. Due to the large number (6) of phosphate groups
Chromatography and desalting of nucleotides with ammonium biocarbonate on anion-exchange columns.
W E COHN et al.
Biochimica et biophysica acta, 48, 588-590 (1961-04-15)
Andrej Shevchenko et al.
Nature protocols, 1(6), 2856-2860 (2007-04-05)
In-gel digestion of proteins isolated by gel electrophoresis is a cornerstone of mass spectrometry (MS)-driven proteomics. The 10-year-old recipe by Shevchenko et al. has been optimized to increase the speed and sensitivity of analysis. The protocol is for the in-gel
Ping Zhang et al.
Molecules (Basel, Switzerland), 23(5) (2018-04-28)
Since the cost of Ophiocordyceps sinensis, an important fungal drug used in Chinese medicine, has increased dramatically, and the counterfeits may have adverse health effects, a rapid and precise marker using the peptide mass spectrometry identification system could significantly enhance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service