Skip to Content
Merck

8.20176

2-Bromoethylammonium bromide

for synthesis

Synonym(s):

2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2H7Br2N
CAS Number:
Molecular Weight:
204.89
UNSPSC Code:
12352107
EC Index Number:
219-924-2
NACRES:
NA.22
MDL number:
Form:
crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

crystals

potency

200-2000 mg/kg LD50, oral (Rat)

pH

2.9 (20 °C, 10 g/L in H2O)

mp

169-176 °C

solubility

>500 g/L

storage temp.

2-30°C

SMILES string

[Br-].BrCC[N+H3]

InChI

1S/C2H6BrN.BrH/c3-1-2-4;/h1-2,4H2;1H

InChI key

WJAXXWSZNSFVNG-UHFFFAOYSA-N

Application

  • Concepts for New Rapid Simple HPLC Method for Quantification of Fosfomycin Trometamol: This study utilized 2-aminoheptane in HPLC methods to quantify fosfomycin trometamol, showcasing its utility in pharmaceutical analysis (M Piponski et al., 2022).

Analysis Note

  • Assay (argentometric): ≥ 99.0 %
  • Melting range (lower value): ≥ 169 °C
  • Melting range (upper value): ≤ 176 °C
  • Identity (IR): passes test


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

142.9 °F - Pensky-Martens closed cup

flash_point_c

61.6 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library