Skip to Content
Merck
All Photos(1)

Documents

Y0001510

Lactulose for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lactulose, 4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
93773
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lactulose

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO)O[C@@](O)(CO)[C@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)7(17)8(18)11(21-4)22-9-5(2-14)23-12(20,3-15)10(9)19/h4-11,13-20H,1-3H2/t4-,5-,6+,7+,8-,9-,10+,11+,12+/m1/s1

InChI key

JCQLYHFGKNRPGE-WJONTELPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lactulose for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heather Lee-Robichaud et al.
The Cochrane database of systematic reviews, (7)(7), CD007570-CD007570 (2010-07-09)
Constipation is a common clinical problem. Lactulose and Polyethylene Glycol (PEG) are both commonly used osmotic laxatives that have been shown to be effective and safe treatments for chronic constipation. However, there is no definitive data as to which provides
Parmjit S Panesar et al.
Biotechnology advances, 29(6), 940-948 (2011-08-23)
Lactulose a "bifidus factor" is composed of galactose and fructose, which can be produced by the isomerization of lactose. It is a prebiotic carbohydrate which stimulates the growth of health-promoting bacteria in the gastrointestinal tract, such as bifidobacteria and lactobacilli
Srinivasan Dasarathy
Indian journal of gastroenterology : official journal of the Indian Society of Gastroenterology, 22 Suppl 2, S50-S53 (2004-03-18)
The pathogenesis of hepatic encephalopathy is mediated to a large extent by ammonia, mainly derived from the gut. Both bacterial and nonbacterial mechanisms of ammoniagenesis have been shown, but ammoniagenesis mediated by colonic bacteria is probably of clinical significance. The
F L Weber
Digestive diseases (Basel, Switzerland), 14 Suppl 1, 53-63 (1996-01-01)
Lactulose is the most frequently utilized agent in the treatment of hepatic encephalopathy because of its efficacy and the fact that it has few serious side effects. How this nonabsorbable disaccharide works has been a matter of controversy, but evidence
B Vogt et al.
Scandinavian journal of gastroenterology. Supplement, 222, 100-101 (1997-01-01)
The introduction of lactulose as a new therapeutic agent for treatment of hepatic encephalopathy was a major breakthrough in this field. It was hypothesized that lactulose might prevent postoperative renal impairment after biliary surgery in patients with obstructive jaundice. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service