Skip to Content
Merck

Skip To

F0756

Fusidic acid

Antibiotic

Synonym(s):

Ramycin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C31H48O6
CAS Number:
Molecular Weight:
516.71
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352106
EC Number:
230-256-0
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fusidic acid,

Quality Segment

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@@]3(C)[C@@]([H])([C@H](O)C[C@@]4([H])\C([C@H](C[C@]34C)OC(C)=O)=C(/CC\C=C(\C)C)C(O)=O)[C@@]1(C)CC[C@@H](O)[C@H]2C

InChI

1S/C31H48O6/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36)/b26-20-/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-/m0/s1

InChI key

IECPWNUMDGFDKC-MZJAQBGESA-N

Application

Fusidic acid has been used:
  • as a standard in thin-layer chromatography (TLC)-densitometric method
  • to study its ability to bind to human serum albumin
  • as an antibiotic in combination with tannic acid to determine minimum inhibitory concentrations (MIC) in methicillin-resistant Staphylococcus aureus (MRSA)

Biochem/physiol Actions

Fusidic acid is a tetracyclic triterpenoid with antibiotic activity against Gram-positive bacteria. It is derived from Fusidium coccineum. It shows its activity against anaerobes, corynebacterial, Nocardia, and Neisseria species. Fusidic acid exhibits therapeutic effects against Staphylococcal infections, skin infections.
Suppresses nitric oxide lysis of pancreatic islet cells. Inhibits protein synthesis in prokaryotes by inhibiting the ribosome-dependent activity of G factor and translocation of peptidyl-tRNA.
Suppresses nitric oxide lysis of pancreatic islet cells; inhibits protein synthesis in prokaryotes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0881I1656C7698
description

-

description

≥98% (TLC)

description

solid

description

from microbial, ≥94% (TLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Fusidic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters