Skip to Content
Merck
All Photos(1)

Key Documents

Y0001362

Nicergoline for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Nicergoline, 5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrN3O3
CAS Number:
Molecular Weight:
484.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nicergoline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12Cc3cn(C)c4cccc(c34)[C@]1(C[C@@H](COC(=O)c5cncc(Br)c5)CN2C)OC

InChI

1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1

InChI key

YSEXMKHXIOCEJA-FVFQAYNVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicergoline for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Young Kim et al.
Current eye research, 36(1), 29-33 (2010-12-23)
We wanted to investigate the effect of nicergoline on corneal nerve regeneration in rat corneas after photorefractive keratectomy (PRK). Twenty Sprague-Dawley male rats were divided into two groups, the control group and the group that had been treated with nicergoline
[A case of pseudotumorous course of brain demyelinating disease].
Iu V Kislitsyn et al.
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova, 105(3), 61-63 (2005-04-13)
Jun-Sub Choi et al.
The ocular surface, 11(1), 16-18 (2013-01-17)
Many factors are involved in the corneal wound healing mechanism, including adhesion, migration, and proliferation of corneal epithelial cells. Abnormal corneal wound healing leads to corneal edema, neovascularization, scar formation, and poor vision. Three agents, 17β-estradiol, nicergoline, and β-glucan, have
Tetsuya Mizuno et al.
Brain research, 1066(1-2), 78-85 (2005-12-06)
We examined the neuroprotective role of nicergoline in neuron-microglia or neuron-astrocytes co-cultures. Nicergoline, an ergoline derivative, significantly suppressed the neuronal cell death induced by co-culture with activated microglia or astrocytes stimulated with lipopolysaccharide (LPS) and interferon (IFN)-gamma. To elucidate the
V Vitton et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 18(11), 1031-1040 (2006-10-17)
A pontine centre located near the micturition centre controlling external anal sphincter (EAS) motility via noradrenergic neurones has been described in cats. The aim of this study was to determine (i) whether a similar centre controls EAS motility in humans

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service