Skip to Content
Merck
All Photos(1)

Key Documents

N7889

Supelco

Nicergoline

analytical standard, for drug analysis

Synonym(s):

5-Bromonicotinic acid 10-methoxy-1,6-dimethylergoline-8-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26BrN3O3
CAS Number:
Molecular Weight:
484.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12Cc3cn(C)c4cccc(c34)[C@]1(C[C@@H](COC(=O)c5cncc(Br)c5)CN2C)OC

InChI

1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1

InChI key

YSEXMKHXIOCEJA-FVFQAYNVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicergoline is an ergot derivative, which may be used in the treatment of affective, cognitive and behavioral disorders seen in older people. It is an α -adrenergic antagonist that may be used for companion animals like cats and dogs to treat, behavioral disorders, problems with aging, excessive vocaliztion and restessness.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Cerebral and peripheral vasodilator

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maddison.EJ, et al.
Small Animal Clinical Pharmacology (2008)
Nicergoline for dementia and other age associated forms of cognitive impairment
MF and LF
Cochrane database of systematic reviews (Online), 2001-2001 null
Tetsuya Mizuno et al.
Brain research, 1066(1-2), 78-85 (2005-12-06)
We examined the neuroprotective role of nicergoline in neuron-microglia or neuron-astrocytes co-cultures. Nicergoline, an ergoline derivative, significantly suppressed the neuronal cell death induced by co-culture with activated microglia or astrocytes stimulated with lipopolysaccharide (LPS) and interferon (IFN)-gamma. To elucidate the
V Vitton et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 18(11), 1031-1040 (2006-10-17)
A pontine centre located near the micturition centre controlling external anal sphincter (EAS) motility via noradrenergic neurones has been described in cats. The aim of this study was to determine (i) whether a similar centre controls EAS motility in humans
Mohamed I Walash et al.
Journal of AOAC International, 91(2), 349-359 (2008-05-15)
A rapid, simple, and highly sensitive second-derivative synchronous fluorometric method has been developed for the simultaneous analysis of binary mixtures of cinnarizine (CN) and nicergoline (NIC). The method is based upon measurement of the native fluorescence of these drugs at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service