Skip to Content
Merck
All Photos(1)

Key Documents

Y0001062

Losartan potassium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Butyl-4-chloro-1-{[2′-(1H-tetrazol-5-yl)(1,1′-biphenyl)-4-yl]methyl}-1H-imidazole-5-methanol monopotassium salt, 2-butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]- 1H-Imidazole-5-methanol potassium salt, DuP 753, MK 954, Potassium 5-(4′-((2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl)methyl)-[1,1′-biphenyl]-2-yl)tetrazol-1-ide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22ClKN6O
CAS Number:
Molecular Weight:
461.00
Beilstein:
5845770
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

losartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)-c3ccccc3-c4nnnn4[K]

InChI

1S/C22H22ClN6O.K/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22;/h4-7,9-12,30H,2-3,8,13-14H2,1H3;/q-1;+1

InChI key

OXCMYAYHXIHQOA-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Losartan potassium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Losartan potassium is an orally available, potent, and selective competitive angiotensin II receptor type 1 (AT1) antagonist. Losartan potassium is used for treatment of hypertension.
orally available, potent, and selective competitive angiotensin II receptor type 1 (AT1) antagonist

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Terada et al.
The Journal of bone and joint surgery. American volume, 95(11), 980-988 (2013-06-20)
Muscle contusions are a common type of muscle injury and are frequently encountered in athletes and military personnel. Although these injuries are capable of healing in most instances, incomplete functional recovery often occurs because of the development of fibrosis in
Christiane Pees et al.
The American journal of cardiology, 112(9), 1477-1483 (2013-07-23)
Since 2008, when angiotensin II type I receptor blockade with losartan was introduced in the prevention of cardiovascular manifestation of Marfan syndrome (MFS), a specific treatment to address the cardiovascular lesions became available. The present study aimed to compare the
Li-Ping Wang et al.
Biochemical pharmacology, 85(10), 1486-1494 (2013-03-19)
The proliferation of cardiac fibroblasts is implicated in the pathogenesis of myocardial remodeling and fibrosis. Intermediate-conductance calcium-activated K⁺ channels (K(Ca)3.1 channels) have important roles in cell proliferation. However, it is unknown whether angiotensin II (Ang II), a potent profibrotic molecule
K L Goa et al.
Drugs, 51(5), 820-845 (1996-05-01)
Losartan potassium is an orally active, nonpeptide angiotensin II (AII) receptor antagonist. It is the first of a new class of drugs to be introduced for clinical use in hypertension. This novel agent binds competitively and selectively to the AII
L M Burrell
Drug safety, 16(1), 56-65 (1997-01-01)
Losartan potassium is the first of a new class of orally active antihypertensive drugs which antagonise the action of angiotensin (AT) II at the AT1 receptor subtype. Losartan potassium is converted by the liver to the active metabolite E-3174, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service