Skip to Content
Merck
All Photos(1)

Documents

163260

Sigma-Aldrich

4-Chloro-7-nitrobenzofurazan

98%

Synonym(s):

4-Chloro-7-nitro-1,2,3-benzoxadiazole, NBD-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2ClN3O3
CAS Number:
Molecular Weight:
199.55
Beilstein:
614212
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

97-99 °C (lit.)

solubility

chloroform: soluble 50 mg/mL, clear to slightly hazy, faintly yellow to yellow

SMILES string

[O-][N+](=O)c1ccc(Cl)c2nonc12

InChI

1S/C6H2ClN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H

InChI key

IGHBXJSNZCFXNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-7-nitrobenzofurazan (NBD-Cl) is a highly sensitive chromogenic and fluorogenic reagent. It is reported to react spontaneously with trans-1-methoxy-3-(trimethylsilyloxy)-1,3-butadiene (Danishefsky′s diene), to afford regioselectively the silyl enol ether, via normal electron-demand Diels-Alder (NEDDA) reaction. A study of the molecular structure, vibrational spectra and NBO (Natural Bond Orbital) analysis of NBD–Cl has been undertaken. Its utility as a pre-column derivatization agent has been examined.

Application

4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies:
  • Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine
  • Synthesis of functionalized hydroxynaphthofurazan.
  • Spectrophotometric and spectrofluorometric determination of clemastine hydrogen fumarate, loratadine, losartan potassium and ramipril in pharmaceutical formulations.
  • Synthesis of 7-nitrobenzofurazan (NBD)-labeled maleimide, via Diels-Alder reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Molecular structure, vibrational, UV and NBO analysis of 4-chloro-7-nitrobenzofurazan by DFT calculations.
Kurt M, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 79(5), 1162-1170 (2011)
Gholamreza Bahrami et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 850(1-2), 400-404 (2007-01-02)
A new, sensitive and simple high-performance liquid chromatographic method for analysis of topiramate, an antiepileptic agent, using 4-chloro-7-nitrobenzofurazan as pre-column derivatization agent is described. Following liquid-liquid extraction of topiramate and an internal standard (amlodipine) from human serum, derivatization of the
4-Chloro-7-nitrobenzofurazan as a Diels-Alder reagent. A facile access to highly functionalized naphthofurazans.
Vichard D, et al.
Tetrahedron Letters, 42(43), 7571-7574 (2001)
Marco Filice et al.
Organic & biomolecular chemistry, 9(15), 5535-5540 (2011-06-23)
Lipase B from Candida antarctica (CAL-B) has been site-directedly modified by the introduction of a trans,trans-hexadiene moiety onto lipase molecules, identified by MALDI-TOF. This modification on CAL-B permitted its immobilization on Q-Sepharose supports in excellent yields (>95%) when native lipase
Gholamreza Bahrami et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 857(2), 322-326 (2007-08-21)
A sensitive and rapid high-performance liquid chromatographic method for the analysis of fluvoxamine, a selective serotonin reuptake inhibitor in human serum, is described using 4-chloro-7-nitrobenzofurazan as pre-column derivatization agent. The drug and an internal standard (fluoxetine) were extracted from 0.25

Articles

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service