Skip to Content
Merck
All Photos(1)

Key Documents

Y0000877

Bisoprolol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Bisoprolol hemifumarate salt, 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31NO4 · .5C4H4O4
CAS Number:
Molecular Weight:
383.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bisoprolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COc1ccc(COCCOC(C)C)cc1.CC(C)NCC(O)COc2ccc(COCCOC(C)C)cc2

InChI

1S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20H,9-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

VMDFASMUILANOL-WXXKFALUSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bisoprolol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cardioselective β1-adrenoceptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT RE 2

Target Organs

Heart

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Hashiyada et al.
Legal medicine (Tokyo, Japan), 15(2), 103-105 (2012-12-12)
An elderly person died of uncontrolled bradycardia in a hospital. The doctor had prescribed 1.35 mg of bisoprolol fumarate orally, but a nurse mistakenly gave the patient 10 mg of the drug 9 hours prior to her death. Bisoprolol was
Benjamin J Hirsh et al.
Circulation. Heart failure, 5(5), 560-565 (2012-08-03)
Chronotropic incompetence is defined as the inability to reach 80% of heart rate (HR) reserve or 80% of the maximally predicted HR during exercise. The presence of chronotropic incompetence is associated with reduced peak oxygen consumption, and rate-responsive pacing therapy
János Tomcsányi et al.
Orvosi hetilap, 154(7), 267-271 (2013-02-12)
The authors describe two cases of takotsubo cardiomyopathy developing after an abrupt withdrawal of carvedilol and bisoprolol. Takotsubo or stress cardiomyopathy is characterized by acute and reversible cardiac dysfunction without coronary artery disease. It is triggered by acute emotional or
Huichao Chang et al.
Journal of pharmaceutical and biomedical analysis, 71, 104-110 (2012-09-06)
A sensitive, specific liquid chromatography-tandem mass spectrometry (LC-MS/MS) method was established for the quantitative determination of amlodipine and bisoprolol, using clenbuterol as the internal standard (IS). The analytes and IS were isolated from 100μL plasma samples by a simple liquid-liquid
Viktor Nagy
Orvosi hetilap, 154(44), 1731-1734 (2013-10-29)
The prevalence of chronic heart failure in Hungary is 1.6% in the adult population, but it occurs in 15-20% of subjects over 80 years of age. The base of treatment of heart failure is the blockade of the neuro-hormonal system

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service