Skip to Content
Merck
All Photos(1)

Key Documents

Y0000829

Nilutamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5,5-Dimethyl-3-[4-nitro-3-(trifluoromethyl)phenyl]-2,4-imidazolidinedione, Anandron, RU-23908

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F3N3O4
CAS Number:
Molecular Weight:
317.22
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nilutamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)NC(=O)N(c2ccc(c(c2)C(F)(F)F)[N+]([O-])=O)C1=O

InChI

1S/C12H10F3N3O4/c1-11(2)9(19)17(10(20)16-11)6-3-4-8(18(21)22)7(5-6)12(13,14)15/h3-5H,1-2H3,(H,16,20)

InChI key

XWXYUMMDTVBTOU-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nilutamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shan Lu et al.
Molecular cancer therapeutics, 6(7), 2057-2064 (2007-07-11)
The purpose of this study was to determine the effects of 6-amino-2-[2-(4-tert-butyl-phenoxy)-ethylsulfonyl]-1H-pyrimidine-4-one (DL3), a novel synthetic compound with small-molecule drug properties, on androgen-regulated gene expression and cell growth in human prostate cancer cells. LNCaP, 22Rv1, and LAPC-4 cells were used
Ravi A Madan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(14), 4526-4531 (2008-07-17)
We reported previously the first randomized study of any kind in patients with nonmetastatic, castrate-resistant prostate cancer. The study employed vaccine, the hormone nilutamide, and the combined therapy (crossover for each arm) with an endpoint of time to progression. We
Richard Choo et al.
International journal of radiation oncology, biology, physics, 75(4), 983-989 (2009-05-05)
To determine the efficacy of a combined approach of radiotherapy (RT) plus 2-year androgen suppression (AS) as salvage treatment for prostate-specific antigen (PSA) relapse after radical prostatectomy (RP). Seventy-five patients with PSA relapse after RP were treated with salvage RT
Jing Yu et al.
Endocrinology, 151(4), 1822-1828 (2010-03-03)
The mechanisms of testosterone-induced vasodilatation are not fully understood. This study investigated the effect of testosterone on nitric oxide (NO) synthesis and its molecular mechanism using human aortic endothelial cells (HAEC). Testosterone at physiological concentrations (1-100 nm) induced a rapid
Chih-Yang Chang et al.
Fertility and sterility, 85 Suppl 1, 1195-1203 (2006-04-18)
To investigate the effects of androgen and antiandrogen and the expression of androgen receptor on mouse embryonic stem cells (ESCs) and the inner cell mass. Controlled laboratory study. Academic university hospital. Blastocysts from mice developed at the Institute for Cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service