Skip to Content
Merck
All Photos(1)

Documents

Y0000688

Loratadine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester, Loratidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N2O2Cl
CAS Number:
Molecular Weight:
382.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

loratadine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)N1CC\C(CC1)=C2/c3ccc(Cl)cc3CCc4cccnc24

InChI

1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3

InChI key

JCCNYMKQOSZNPW-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Loratadine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-sedating histamine H1-receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

R Wilkinson et al.
Brain injury, 13(12), 1025-1031 (2000-01-11)
The use of neuroleptics in the acute management of traumatic brain injury (TBI) is controversial and may be detrimental to recovery. The following case report describes a patient developing neuroleptic malignant syndrome (NMS) secondary to the use of haloperidol given
Nemanja Kolašinac et al.
International journal of pharmaceutics, 436(1-2), 161-170 (2012-07-10)
The present study investigates the possibility of using poloxamers as solubility and dissolution rate enhancing agents of the poorly water soluble drug substance desloratadine that can be used for the preparation of immediate release tablet formulation. Two commercially available poloxamer
J Kornhuber et al.
Neuropharmacology, 34(7), 713-721 (1995-07-01)
Amantadine (1-amino-adamantane) is clinically used for the management of Parkinson's disease and drug-induced extrapyramidal symptoms. It has previously been shown that amantadine is a low-affinity uncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist with rapid blocking and unblocking channel kinetics (Ki-value at the
H Ebrahimzadeh et al.
Analytica chimica acta, 767, 155-162 (2013-03-05)
In this work a molecularly imprinted polymer was developed as a selective sorbent for extraction of loratadine (as a model) in complex matrices followed by miniaturized homogeneous liquid-liquid extraction (MHLLE) for the first time. The molecularly imprinted polymer (MIP) which
Udo Döbbeling et al.
Experimental dermatology, 22(2), 119-124 (2013-02-01)
Mycosis fungoides and its leukaemic counterpart Sézary syndrome are the most frequent cutaneous T-cell lymphomas (CTCL), and there is no cure for these diseases. We evaluated the effect of clinically approved antihistamines on the growth of CTCL cell lines. CTCL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service