Skip to Content
Merck
All Photos(1)

Key Documents

Y0000603

Loratadine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Loratadine, 4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene-1-piperidinecarboxylic acid ethyl ester, Loratidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23N2O2Cl
CAS Number:
Molecular Weight:
382.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

loratadine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)N1CC\C(CC1)=C2/c3ccc(Cl)cc3CCc4cccnc24

InChI

1S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14H,2,5-6,9-10,12-13H2,1H3

InChI key

JCCNYMKQOSZNPW-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Loratadine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-sedating histamine H1-receptor antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed El-Awady et al.
Journal of chromatography. A, 1309, 64-75 (2013-08-21)
The analysis of hydrophobic basic analytes by micellar electrokinetic chromatography (MEKC) is usually challenging because of the tendency of these analytes to be adsorbed onto the inner capillary wall in addition to the difficulty to separate these compounds as they
D K Strong et al.
DICP : the annals of pharmacotherapy, 25(11), 1175-1177 (1991-11-01)
Amantadine hydrochloride, a dopamine agonist with antiviral and antiparkinsonism properties, is used for the prevention and treatment of influenza A respiratory infections in high-risk populations. The occurrence of amantadine-induced hallucinations and tremors is described in a young, renal transplant patient
M Magerl et al.
The British journal of dermatology, 166(5), 1095-1099 (2012-01-17)
Cold urticaria is a rare but severe and potentially lethal condition. It is primarily treated symptomatically with H(1) -antihistamines. However, patients have a variable response to these drugs and, to date, it has not been possible to predict readily the
P Deep et al.
Synapse (New York, N.Y.), 34(4), 313-318 (1999-10-26)
The efficacy of amantadine in alleviating motor symptoms of Parkinson's disease may be mediated in part by stimulation of cerebral dopa decarboxylase (DDC) activity, secondary to antagonism of N-methyl-D-aspartate (NMDA) type glutamate receptors. We tested the specific hypothesis that amantadine
Karsten Weller et al.
Acta dermato-venereologica, 93(2), 168-174 (2012-10-12)
Non-sedating H1-antihistamines are the recommended first-line treatment for chronic spontaneous urticaria. While efficacy studies usually apply continuous daily treatment regimens, many patients take their medication on demand. In this randomized, double-blind trial we tested whether on-demand H1-antihistamine desloratadine in standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service