Skip to Content
Merck
All Photos(1)

Key Documents

Y0000481

Dibrompropamidine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dibrompropamidine diisetionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30Br2N4O10S2
CAS Number:
Molecular Weight:
722.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dibrompropamidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)CCO.Brc1c(ccc(c1)\C(=N\[H])\N)OCCCOc2c(cc(cc2)\C(=N\[H])\N)Br

InChI

1S/C17H18Br2N4O2.C2H6O4S/c18-12-8-10(16(20)21)2-4-14(12)24-6-1-7-25-15-5-3-11(17(22)23)9-13(15)19;3-1-2-7(4,5)6/h2-5,8-9H,1,6-7H2,(H3,20,21)(H3,22,23);3H,1-2H2,(H,4,5,6)

InChI key

BZACJASHKPPTKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dibrompropamidine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[When the treatment is the problem].
Dag Sollesnes Holsen
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 129(4), 307-307 (2009-02-24)
M T Suller et al.
The Journal of hospital infection, 43(4), 281-291 (2000-02-05)
Concern has been growing regarding the potential of antibiotic and disinfectant co-resistance in clinically important bacteria. In this study, the susceptibilities of methicillin-resistant Staphylococcus aureus (MRSA) and methicillin-sensitive Staphylococcus aureus (MSSA) to chlorhexidine (CHX), the quaternary ammonium compounds cetylpyridinium chloride
D Boillot et al.
Diabetologia, 28(6), 359-364 (1985-06-01)
The antiprotozoal drug pentamidine can be toxic to islet cells in vivo and in vitro. Rat islets were exposed to pentamidine (mesylate and isethionate salts) and six other structurally related diamidines. The beta-cell response to arginine + theophylline was suppressed
Victoria W Y Wong et al.
Eye & contact lens, 33(6 Pt 1), 329-331 (2007-11-13)
To describe a patient with a good visual outcome after prompt treatment of Acanthamoeba keratitis as a complication of overnight orthokeratology lens wear. Interventional case report. A 9-year-old boy experienced pain, photophobia, and redness in his right eye 3 days
R Yeoh et al.
The British journal of ophthalmology, 71(7), 500-503 (1987-07-01)
We report on four patients seen during the past two years who had acanthamoeba keratitis. One is described in detail. We believe that acanthamoebic eye infection is not rare. An immunofluorescent technique for localisation of amoebae and cysts is described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service