Skip to Content
Merck
All Photos(1)

Documents

Y0000396

Granisetron hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N4O· HCl
CAS Number:
Molecular Weight:
348.87
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

granisetron

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN1[C@H]2CCC[C@@H]1C[C@@H](C2)NC(=O)c3nn(C)c4ccccc34

InChI

1S/C18H24N4O.ClH/c1-21-13-6-5-7-14(21)11-12(10-13)19-18(23)17-15-8-3-4-9-16(15)22(2)20-17;/h3-4,8-9,12-14H,5-7,10-11H2,1-2H3,(H,19,23);1H/t12-,13+,14-;

InChI key

QYZRTBKYBJRGJB-WQTKJZBYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Granisetron hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Granisetron hydrochloride is a serotonin 5-HT3 receptor antagonist and antiemetic.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hea-Jeong Doh et al.
Colloids and surfaces. B, Biointerfaces, 101, 475-480 (2012-09-27)
A new lipid nanoemulsion (LNE) system containing granisetron (GRN) was developed and its in vitro permeation-enhancing effect was evaluated using Caco-2 cell monolayers. Particle size, polydispersity index (PI) and stability of the prepared GRN-loaded LNE systems were also characterized. The
Mohammad-Reza Khodaie-Ardakani et al.
Journal of psychiatric research, 47(4), 472-478 (2013-02-05)
Some 5-HT3 antagonists such as ondansetron have shown beneficial effects on negative symptoms of patients with schizophrenia. We aimed to evaluate the efficacy of granisetron (another 5-HT3 antagonist) add-on therapy in the treatment of negative symptoms of patients with stable
Joseph A Roscoe et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 30(27), 3389-3395 (2012-08-24)
We conducted a double-blind randomized clinical trial of the following four regimens for controlling delayed nausea (DN): group 1: palonosetron + dexamethasone on day 1 with prochlorperazine on days 2 and 3; group 2: granisetron + dexamethasone on day 1
J Vardy et al.
Breast cancer research and treatment, 136(1), 143-151 (2012-09-08)
Uncertainty remains about the optimal anti-emetic regimen for control of delayed nausea and vomiting after adjuvant chemotherapy for breast cancer. Many patients receive dexamethasone but complain of insomnia, anxiety/agitation, and indigestion. The aim was to determine if patients receiving chemotherapy
Jennifer Fazzari et al.
International journal of radiation biology, 88(10), 786-790 (2012-07-25)
To test the importance of serotonin as a signaling molecule involved in the production and response of radiation-induced bystander effects. HPV-G human keratinocyte cultures were spiked with various concentrations of Granisetron or Ketanserin and subject to either 0 Gy or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service