Skip to Content
Merck
All Photos(3)

Key Documents

S0929

Sigma-Aldrich

Scopolamine hydrobromide

meets USP testing specifications

Synonym(s):

(−)-Scopolamine hydrobromide trihydrate, Hyoscine hydrobromide, Scopine tropate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HBr · 3H2O
CAS Number:
Molecular Weight:
438.31
Beilstein:
6100669
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

mp

195-199 °C (dry matter) (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.Br.CN1[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C17H21NO4.BrH.3H2O/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;;;;/h2-6,11-16,19H,7-9H2,1H3;1H;3*1H2/t11-,12-,13-,14+,15-,16+;;;;/m1..../s1

InChI key

LACQPOBCQQPVIT-SEYKEWMNSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Scopolamine hydrobromide has been used as a muscarinic receptor antagonist:
  • to reverse the cardiac effects of physostigmine in rabbit heart
  • to study its effect on acoustic startle reflex (gap-PPI){148)
  • to study it′s in vitro antiviral, antibacterial, and antifungal activities and cytotoxicity
  • to study its effect on conditioning of lever pressing

Biochem/physiol Actions

Competitive nonselective muscarinic acetylcholine antagonist.
Competitive nonselective muscarinic acetylcholine antagonist. Scopolamine-induced amnesia in laboratory animals is a commonly-used model of memory deficit.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth R I Barker et al.
Learning & memory (Cold Spring Harbor, N.Y.), 16(1), 8-11 (2009-01-02)
Object-in-place memory, which relies on the formation of associations between an object and the place in which it was encountered, depends upon a neural circuit comprising the perirhinal (PRH) and medial prefrontal (mPFC) cortices. This study examined the contribution of
Reversal of cardiac vagal effects of physostigmine by adjunctive muscarinic blockade
Winter J, et al.
Neurotoxicology, 57, 174-182 (2016)
Effects of Scopolamine on Conditioning of Lever Pressing
Juarez Y, et al.
Acta Acad. Med. Sin., 1(1), 14-23 (2011)
Bardia Nouriziabari et al.
Journal of Alzheimer's disease : JAD, 63(2), 725-740 (2018-04-18)
Trace eyeblink conditioning is a hippocampus-dependent associative learning paradigm which is impaired in patients with Alzheimer's disease (AD) and animal AD models. Learning in this paradigm accompanies changes in oscillatory activity in forebrain regions, some of which are loci of
Patrizia Giannoni et al.
The Journal of pharmacology and experimental therapeutics, 332(1), 164-172 (2009-10-10)
After oral administration, the nonimidazole histamine H(3) receptor antagonist, 6-[(3-cyclobutyl-2,3,4,5-tetrahydro-1H-3-benzazepin-7-yl)oxy]-N-methyl-3-pyridinecarboxamide hydrochloride (GSK189254), increased histamine release from the tuberomammillary nucleus, where all histaminergic somata are localized, and from where their axons project to the entire brain. To further understand functional histaminergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service