Skip to Content
Merck
All Photos(1)

Key Documents

PHR3668

Supelco

Losartan Impurity G

certified reference material, pharmaceutical secondary standard

Synonym(s):

Triphenylmethanol, Triphenylcarbinol, Trityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3COH
CAS Number:
Molecular Weight:
260.33
Beilstein:
1460837
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
pharmaceutical secondary standard

Quality Level

API family

losartan

packaging

pkg of 30MG

bp

360 °C (lit.)

mp

160-163 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

OC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H16O/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,20H

InChI key

LZTRCELOJRDYMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service