Skip to Content
Merck
All Photos(1)

Key Documents

PHR1588

Supelco

Piroxicam

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Piroxicam, 4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P2130000
traceable to USP 1544508

API family

piroxicam

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)

InChI key

QYSPLQLAKJAUJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Piroxicam belongs to the class of acidic, non-steroid anti-inflammatory (NSAI) drugs, with a long half-life which enables it to be administered once daily.

Application

Piroxicam may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and urine by spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Cyclooxygenase inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA4573 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anal. Profiles Drug Subst., 15, 509-531 (1986)
A simple spectrofluorimetric method for determination of piroxicam and propranolol in pharmaceutical preparations.
Tabrizi AB
Journal of food and drug analysis, 15(3) (2007)
Determination of piroxicam in pharmaceutical formulations and urine samples using europium-sensitized luminescence.
Al-Kindy SMZ, et al.
Journal of Luminescence, 127(2), 291-296 (2007)
Spectrophotometric determination of piroxicam and tenoxicam in pharmaceutical formulations using alizarin.
Amin AS
Journal of Pharmaceutical and Biomedical Analysis, 29(4), 729-736 (2002)
Sensitive spectrophotometric methods for the determination of amoxycillin, ciprofloxacin and piroxicam in pure and pharmaceutical formulations.
Nagaralli BS, et al.
Journal of Pharmaceutical and Biomedical Analysis, 29(5), 859-864 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service