PHR2183
Diisopropyl Ether
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Diisopropyl ether, Isopropyl ether
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
vapor density
3.5 (vs air)
vapor pressure
120 mmHg ( 20 °C)
CofA
current certificate can be downloaded
autoignition temp.
827 °F
expl. lim.
1-21 %, 100 °F
packaging
ampule of 3x1.2 mL
refractive index
n20/D 1.367 (lit.)
bp
68-69 °C (lit.)
mp
−85 °C (lit.)
density
0.725 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-30°C
SMILES string
CC(C)OC(C)C
InChI
1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3
InChI key
ZAFNJMIOTHYJRJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Recommended products
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-20.2 °F - closed cup
Flash Point(C)
-29 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service