Skip to Content
Merck
All Photos(5)

Key Documents

1.00867

Supelco

Diisopropyl ether

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

2,2′-Oxybis[propane], 2,2-Propoxypropane, Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-560-6
NACRES:
NA.21
grade:
ACS reagent
for analytical purposes
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. Ph. Eur.

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

product line

EMSURE®

Assay

≥99% (GC)

form

liquid

autoignition temp.

827 °F

potency

5880 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

contains

~10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

expl. lim.

1-21 %, 100 °F

dilution

(for analytical testing)

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Titrable acid
≤0.001% Peroxide (as H2O2)

evapn. residue

≤0.005%

color

clearAPHA: ≤10

refractive index

n20/D 1.367 (lit.)

pH

7 (20 °C)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

transition temp

flash point -28 °C

density

0.725 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CC(C)OC(C)C

storage temp.

2-30°C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service