Skip to Content
Merck
All Photos(1)

Key Documents

PHR1093

Supelco

Sulfanilamide melting point standard

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sulfanilamide, p-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
39093202
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1633007

API family

sulfanilamide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

~Approximately 166 °C
164-166 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sulfanilamide Melting Point Standard, also called Sulfanilamide, is an antibacterial drug. It blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase.
Sulfanilamide Melting Point Standard also called Sulfanilamide is an antibacterial drug.

Application

It was used in a study to demonstrate photodecomposition in aqueous solution of cutaneous photosensitizing agents with the help of spin traps 5, 5-dimethyl-1-pyrroline-1-oxide.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB1410 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SPECTROSCOPIC STUDIES OF CUTANEOUS PHOTOSENSITIZING AGENTS?II. SPIN TRAPPING OF PHOTOLYSIS PRODUCTS FROM SULFANILAMIDE AND 4-AMINOBENZOIC ACID USING 5, 5-DIMETHYL-1-PYRROLINE-1-OXIDE.
Chignell, Colin F., et al.
Photochemistry and Photobiology, 34.2 , 147-156 (1981)
Sevgi Kolayli et al.
Journal of enzyme inhibition and medicinal chemistry, 26(6), 895-900 (2011-03-09)
An α-carbonic anhydrase (CA, EC 4.2.1.1) was purified and characterized kinetically from erythrocytes of the sturgeon Acipenser gueldenstaedti, an endangered species. The sturgeon enzyme (AgCA) showed kinetic parameters for the CO(2) hydration reaction comparable with those of the human erythrocytes
Penghua Wang et al.
Water research, 45(16), 5015-5026 (2011-08-02)
A novel carbon-sensitized and nitrogen-doped TiO2 (C/N-TiO2) was synthesized by a facile sol-gel method using titanium butoxide as both titanium precursor and carbon source, and nitric acid as nitrogen source. The calcination temperature had a great effect on the crystal
Ramazan Demirdağ et al.
Bioorganic & medicinal chemistry, 21(6), 1522-1525 (2012-09-15)
Carbonic anhydrase (CA, EC: 4.2.1.1) was purified from sheep kidney by affinity chromatography on a Sepharose 4B-tyrosine-sulfanilamide column. By means of two consecutive procedures, the enzyme (sCA) was purified 227.61-fold with a yield of 60.75%, and a specific activity of
Fei Liu et al.
Organic letters, 12(4), 868-871 (2010-01-19)
The synthesis of benzofused sultams and/or fluorinated sulfonamides, starting from N-allylic sulfonamides, was performed in superacid HF/SbF(5), through superelectrophilic activation. A dramatic effect of superacid composition on the selectivity of the reaction was shown and applied to the synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service