Skip to Content
Merck
All Photos(1)

Key Documents

PHR1137

Supelco

Isopropyl palmitate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Isopropyl hexadecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOCH(CH3)2
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786567
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. I0725000
traceable to USP 1350603

API family

isopropyl palmitate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.438 (lit.)

mp

11-13 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C19H38O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19(20)21-18(2)3/h18H,4-17H2,1-3H3

InChI key

XUGNVMKQXJXZCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropyl palmitate is an analog of isopropyl myristate and an aliphatic ester used as a flavoring ingredient in food industry. It is one of the volatile compounds found in Psidium salutare fruits and boiled buckwheat flour.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7156 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Food Drug and Cosmetic Excipients
CRC Press (1992)
Varaporn Buraphacheep Junyaprasert et al.
Drug delivery, 15(5), 323-330 (2008-09-03)
The skin permeation enhancement of many kinds of drugs and cosmetic substances by microemulsions has been widely known; however, the correlations between microemulsion microstructures and the efficiency of skin permeation are not fully elucidated. Therefore, the aim of our study
Honglei Xi et al.
International journal of pharmaceutics, 436(1-2), 857-861 (2012-07-17)
The aim of this work was to investigate the effect of the stability of hydrogen-bonded ion pairs with organic amines on transdermal penetration of teriflunomide (TEF). Five organic amines, diethylamine (DEtA), triethylamine (TEtA), diethanolamine (DEA), triethanolamine (TEA), and N-(2'-hydroxyethanol)-piperdine (NP)
G Häring et al.
Biochemical and biophysical research communications, 127(3), 911-915 (1985-03-29)
A reverse micellar system containing Tween 85 and water in isopropylpalmitate was developed which permitted the solubilization of bacteria in the form of homogenous organic solutions. The presence of the bacteria in solution was demonstrated by light microscopy. Immediately after
N Pfammatter et al.
Biochemical and biophysical research communications, 161(3), 1244-1251 (1989-06-30)
Baker yeast cells are solubilized in organic solvents by the use of surfactants and small amounts of water. Data are reported for three different systems, Tween/Isopropylpalmitat, Asolectin/IPP, Asolectin/Hexadecane. The viability (life-capability) can remain as high as 80% for 10 Days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service